Millicup™-FLEX Disposable Vacuum Filtration Units and demo video
Millicup™-FLEX disposable vacuum filtration units place control in your hands, providing the benefits of traditional glassware with the time-saving flexibility of disposable solutions.
Connect the threaded membrane support collar to a vacuum-rated, receiver bottle by turning in a clockwise manner until the collar is fully tightened, ensuring the gasket remains in place.
Using smooth-tip forceps, place a 47 mm membrane disc onto the support collar.
Without disturbing the filter, center the base of the funnel on top of the membrane support base and gently press down until you hear an audible “click”.
Connect vacuum source and filter solution
Attach one of the included tube adapters to the integrated vacuum port, connecting one end of the vacuum tubing to the system and the other end to the vacuum source.
Pour sample into funnel and apply vacuum until filtration is complete, rinsing the funnel (if required) while the vacuum is on.
Remove filtration device and recycle
Turn off the vacuum and remove tubing. If membrane removal is desired, gently lift up on the funnel for access.
Remove the collar by turning in a counter-clockwise manner until loosened. If desired, place a cap onto the receiver bottle for storage.
How to Recycle Millicup™-FLEX Disposable Vacuum Filter Units
All three components of the Millicup™-FLEX unit are fully recyclable, minimizing your environmental impact. Before placing the unit in your lab’s mixed plastic or single-stream recycling, you must remove the membrane and rinse any residual solids from the funnel and support collar.