Mobilize Your Laboratory with Our Spectroquant® Move Instruments
A robust portable handheld colorimeter for rapid on-site analyses with reliable results
No matter what you need to test - our Spectroquant® Move instruments offer dependable results for a broad range of pre-programmed parameters with an independent instrument, wherever you need it.
No matter where you need to test – our Spectroquant® Move colorimeters are robust, dust and waterproof according to IP 68.
No matter where you need your data – the Spectroquant® Data Transfer Unit allows you to transfer, print & store your measurements.
Move 100 – for Water Analysis
  • Rapid, reliable on-site water analysis with a compact portable colorimeter
  • Move 100 offers over 100 pre-programmed parameters
Technical Data
Scope of Delivery: Instrument in light carrying case, 4 batteries, 3 round vials each 24- and 16-mm ø, 1 adapter for 16-mm vials, screw driver, guarantee certificate, certificate of compliance, instruction manual
Display: Backlit graphic-display
Interfaces: IR interface for data transfer & RJ45 connector for internet updates
Optics: LED interference filter, photosensor, transparent measurement chamber
Wavelength: 430, 530, 560, 580, 610 and 660 nm
Wavelength accuracy: ± 1nm
Photometric accuracy:
1.000 Abs ± 0.020 Abs
2.600 Abs ± 0.052 Abs (2 % FS)
(measured with standard solution – T = 20 – 25°C)
Photometric resolution:  0.005 A
Operation: Acid and solvent resistant tactile film keyboard
Power supply: 4 batteries (Type AA / LR6), capacity approx. 26 hours continuous use or 3,500 tests
Weight: Approx. 450 g
Dimensions:
Approx. 210 x 95 x 45 mm (Instrument)
Approx. 395 x 295 x 106 mm (Case)
IP classification:  Dust and waterproof acc. to IP 68
Storage capacity:  approx. 1,000 data sets
Move DC – for Disinfection Control
  • Reliable results for the key disinfection parameters (Cl2/O3/ClO2/CyA/pH) with validated standard-compliant reagents.
  • Comprehensive range of Spectroquant® test kits to measure disinfectants before and after your cleaning process
Technical Data
Scope of Delivery: Instrument in light carrying case, 4 batteries, 3 round vials each 24- and 16-mm ø, 1 adapter for 16-mm vials, screw driver, guarantee certificate, certificate of compliance, instruction manual
Display: Backlit LCD (on keypress)
Interfaces: IR interface for data transfer
Optics: LED interference filter, photosensor, transparent measurement chamber
Wavelength: 530 nm
Wavelength accuracy: ± 1nm
Photometric accuracy:
1.000 Abs ± 0.030 Abs
2.600 Abs ± 0.078 Abs (3 % FS)
(measured with standard solution – T = 20 – 25°C)
Photometric resolution:  0.001 A
Operation: Acid and solvent resistant tactile film keyboard
Power supply: 4 batteries (Type AAA / LR03), capacity approx. 17 hours continuous use or 5,000 tests
Weight: Approx. 260 g
Dimensions:
Approx. 155 x 75 x 35 mm (Instrument)
Approx. 340 x 275 x 83 mm (Case)
IP classification:  Dust and waterproof acc. to IP 68
Storage capacity: Internal ring memory for 16 data sets
Materials