GLP-Plates
With Individual Laser Coding for Secure Documentation
GLP-Plates
Based on the proven silica gel 60, manufactured by Merck KGaA, Darmstadt, Germany, our GLP plates deliver the same unsurpassed separation performance as the corresponding TLC or HPTLC plates. The only difference is that each of our laser-coded GLP plates is marked with an item, batch, and individual plate number. This allows you to easily record, document and archive every plate you use. The GLP plates are available as TLC or HPTLC grade in various formats, with or without the fluorescence indicator F254, which is stimulated to green emission at 254 nm.
Features and benefits
  • Convenient back tracing of article, batch, and individual plate number
  • Easy documentation and archiving of every plate
  • Same reliable performance as our TLC and HPTLC plates
TLC Plates GLP
Product No. Description Layer Sorbent/Coating Material Layer Thickness/ µm Format /cm Support/ Backing Pack Size / Content
1.05566.0001 TLC silica gel 60 F254 GLP Silica gel 60 F254*1 250 20 x 20 glass 25 plates
1.05702.0001 TLC silica gel 60 F254 GLP Silica gel 60 F254*1 250 10 x 20 glass 25 plates
*1 F254 Fluorescent indicator
HPTLC Plates GLP
Product No. Description Layer Sorbent/Coating Material Layer Thickness/ µm Format /cm Support/ Backing Pack Size / Content
1.13326.0001 HPTLC silica gel 60 GLP Silica gel 60 250 20 x 10 glass 25 plates
1.05702.0001 HPTLC silica gel 60 F254 GLP Silica gel 60 F254* 250 20 x 10 glass 25 plates
1.05564.0001 HPTLC silica gel 60 F254 GLP Silica gel 60 F254*1 250 10 x 10 glass 25 plates
*1 F254 Fluorescent indicator