Aluminum Oxide TLC Plates
Excellent Separations of Basic and Neutral Compounds
Our aluminum oxide (alox) plates for thin-layer chromatography offer distinct advantages with regard to pH. Under aqueous conditions, basic compounds can be best separated on basic alox plates, while neutral alox plates are most suitable for neutral compounds. The plates achieve excellent separations for a wide variety of applications thanks to neutral and basic aluminum oxides with 60Å pore sizes.
TLC Aluminum Oxide 60
Product No. Description Layer Sorbent/ Coating Material Layer thickness/ µm Format/ cm Support/ Backing Pack size/ Content
1.05713.0001 TLC Aluminum oxide 60 F254  ,basic Aluminum oxide 60 F254 *1 basic 250 20 x 20 glass 25 plates
1.05731.0001 TLC Aluminum oxide 60 F254 *   ,basic Aluminum oxide 60 F254 *1 basic 250 5 x 20 glass 100 plates
1.05550.0001 TLC  Aluminum oxide 60 F254  , neutral Aluminum oxide 60 F254 *1 neutral 200 20 x 20 aluminum 25 sheets
1.05581.0001 TLC Aluminum oxide 60 F254  , neutral Aluminum oxide 60 F254 *1 neutral 200 20 x 20 plastic 25 sheets
*1 F254 Fluorescent indicator