High-Resolution U/HPLC Separation of Peptides BIOshell™ A160 Peptide Phenyl-Hexyl
High efficiency separation of peptides with orthogonal selectivity to alkyl-bonded phases due to rigid aromatic ring (shape selectivity).
The BIOshell™ A160 Peptide Phenyl-Hexyl bonded phase is based on fused-core particles. BIOshell™ A160 Peptide Phenyl-Hexyl provides a stable, reversed phase packing with a pore structure and pore size that is optimized for reversed-phase HPLC separations of peptides and polypeptides, using typical acidic mobile phases favored for protein structure-function and proteomic applications. The aromaticity of the stationary phase ligand can interact with amino acids with aromatic side chains, allowing for differential selectivity not afforded by alkyl-bonded phases.
Having a phenyl-hexyl bonding, methods developed on this column should be screened with both methanol and acetonitrile, supplemented with an ion-pairing reagent like 0.1% (v/v) trifluoroacetic acid (TFA) or difluoroacetic acid (DFA), as these two solvents have a wider eluotropic strength with stationary phases with aromaticity than is seen with alkyl-bonded phases. For typical peptide workflows, column temperatures of 40 – 60 °C are used; higher temperatures can enhance peak shape and can increase the throughput of the workflow.
BIOshell™ A160 Peptide Phenyl-Hexyl Specifications:
Silica: Type B (High purity silica)
Particle Platform: Superficially porous particles (SPP)
Phase Chemistry: Dimethylphenyl-hexyl
Particle Size: 2.7 µm
Pore Size: 160 Å
Carbon Load: 4.7 %
Surface Area: 90 m²/g
pH Range: 2 - 9
Max Temperature at Low pH: 90 °C
Max Temperature at High pH: 40 °C
Endcapped: Yes
BIOshell™ A160 Peptide Phenyl-Hexyl is a high-speed, high-performance liquid chromatography column based on a superficially porous Fused-Core® particle design. The Fused-Core® particle provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths.
BIOshell™ A160 Peptide Phenyl-Hexyl U/HPLC Columns, 2.7 µm
Outstanding performance in U/HPLC. 2.7 µm SPP particle with 160 Å pores. Ideal for the separation of peptides
2.7 µm SPP particle with 1000 Å pores