Use this interactive biological buffers selection guide below to help find the right buffer by pH, grade, packaging type, or application.
Refer to the Buffer Reference Center to prepare specific buffer solutions by pH and pKa.
Try pHast Pack™ for free – New Ready-to-Use Buffers
Watch the Webinar: Getting Your Buffers Right: How to Control the pH of Cell Culture Medium
![]() |
Packaging Options |
• Convenience Packaging • Redi-Dri™ free-flowing powders • Solutions • Capsules • Tablets • Preweighed Powders • Bulk Powder Transfer Bags |
![]() |
Learning Center |
• Buffer Reference Center • Request your Buffer Selection Guide (Printed Copy | PDF) • Information on Expiration and Retest Dating • Reproducibility with Biological Buffers |
By definition, biological buffers are organic substances that maintain a constant pH over a given range by neutralizing the effects of hydrogen ions. In the body, buffers provide a pH environment conducive to critical biochemical processes.
Buffers are ubiquitous in modern biology labs, especially in applications involving protein electrophoresis to view the results of a technique such as the polymerase chain reaction or Western blotting. They are a vital component for modeling biological systems and have many uses in cell culture, molecular biology, nucleic acid and protein purification, and transformation and transfection.
Two common chemistry buffer examples are tris(hydroxymethyl)aminomethane (tris) and 4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid (HEPES), and many others are listed below.
A buffering range or buffer range indicates the pH level that a buffer maintains over the course of an experiment.
Buffers are typically classified into seven main groups based on the R-group in their chemical makeup and the pH range of their effectiveness. They can be further delineated by grade. At the acidic end of the pH scale, at a range from 5.5 to 8.3 are the morpholinic buffers such as MES, MOPS, and MOBS. Tris is a mid-range buffer stable at pH of 7 to 9.6. Cyclohexylamino buffers such as CHES and CAPS are effective at higher pH values from 8.6 to 11.4.