Agarose Selection By Application
Sort the table by clicking in the table headers. To reset the table, click here.
Product / No.                                   Analytical Separation of DNA ≥ 1000 bp    Analytical Separation of DNA ≤ 1000 bp   
Pulsed Field Gel Electrophoresis Fine
Resolution
Enzyme
Activity
Protein Electrophoresis DNA
Extraction
Immunoelectrophoresis
and Immunodiffusion
Heat Labile Materials
Agarose / A4679
Low EEO
              x  
Agarose / A9311
Medium EEO
              x  
Agarose / A5304
Special high EEO
              x  
Agarose / A9539
BioReagent, for molecular biology, low EEO
x x         x    
Agarose / A0701
Type VII-A
        x x x   x
Agarose/ A4018
Low gelling temperature
        x   x   x
Agarose / A2576
Ultra-low gelling temperature, molecular biology grade
        x   x   x
Agarose / A5030
Ultra-low gelling temperature
        x       x
Agarose / A2929
For pulsed field electrophoresis running gel
x   x       x    
Agarose / A3054
For pulsed field electrophoresis sample preparation
    x       x    
Agarose / A9414
BioReagent, for molecular biology
  x              
Agarose / A2790
Wide range, for molecular biology
x                
Agarose / A7431
BioReagent, for molecular biology, Wide range/Standard 3:1
  x   x         x
Agarose / A9045
BioReagent, suitable for cell culture, insect cell culture & plant cell culture
              x  
Agarose / 05066
High EEO, for molecular biology
            x    
Agarose / A4718
High resolution, for molecular biology
  x   x          
Agarose / A0169
Type I-A, low EEO
x x         x    
Agarose / A0576
Low EEO
x                
Agarose / A6013
Type I, low EEO
x x         x    
Agarose / A9918
Type II-A, Medium EEO
              x  
Agarose / A6877
Medium EEO, for molecular biology
x x           x  
Agarose / A9793
High EEO
              x  
Agarose / A3643
Type IV, special high EEO
          x   x  
Agarose / A7174
High gelling temperature
  x           x  
Agarose LE / AGRLE-RO
Low electroendosmosis
x x         x    
Agarose MP / AGRMP-RO
Multipurpose agarose
  x x       x    
Agarose MS / AGRMS-RO
Molecular screening agarose
x x   x