LOCUS p63 14577 bp DNA circular 22-AUG-2017 DEFINITION . ACCESSION urn.local...9y-7o8qajg KEYWORDS . SOURCE ORGANISM . FEATURES Location/Qualifiers misc_feature 1..6 /Recognition_pattern="C^YCGRG" /label="AvaI" /note="Geneious type: restriction site" misc_feature 179..973 /label="Kan" misc_feature 739..744 /Recognition_pattern="C^CATGG" /label="NcoI" /note="Geneious type: restriction site" misc_feature 1273..1946 /label="pUC Ori" misc_feature 1629..1634 /Recognition_pattern="G^TGCAC" /label="ApaLI" /note="Geneious type: restriction site" misc_feature 1981..1986 /label="SalI" misc_feature 1987..5692 /label="pVS1 Ori" misc_feature 3663..3668 /Recognition_pattern="C^YCGRG" /label="AvaI" /note="Geneious type: restriction site" misc_feature 4143..4148 /Recognition_pattern="C^YCGRG" /label="AvaI" /note="Geneious type: restriction site" misc_feature 4967..4972 /Recognition_pattern="AT^CGAT" /label="ClaI" /note="Geneious type: restriction site" misc_feature 5693..5698 /label="XbaI" misc_feature 5725..5732 /label="PmeI" misc_feature complement(5733..6046) /label="NOS terminator" misc_feature 6003..6008 /Recognition_pattern="C^YCGRG" /label="AvaI" /note="Geneious type: restriction site" misc_feature complement(6047..8050) /label="GUS" misc_feature 8055..8060 /label="Acc65I" misc_feature complement(8061..8891) /label="35S" misc_feature 8892..8897 /Recognition_pattern="G^AATTC" /label="EcoRI" /note="Geneious type: restriction site" misc_feature 8898..9206 /label="U6" allele 9207..9213 /label="AarI" misc_feature 9224..9230 /label="AarI" misc_feature 9235..9317 /label="sgRNA scaffold" misc_feature 9319..9324 /Recognition_pattern="A^AGCTT" /label="HindIII" /note="Geneious type: restriction site" misc_feature 9353..10133 /label="35S" misc_feature 10134..10153 /label="T7" misc_feature 10176..14306 /label="Cas9" misc_feature 11262..11267 /Recognition_pattern="AT^CGAT" /label="ClaI" /note="Geneious type: restriction site" misc_feature 12360..12365 /Recognition_pattern="G^GATCC" /label="BamHI" /note="Geneious type: restriction site" misc_feature 12726..12731 /Recognition_pattern="AT^CGAT" /label="ClaI" /note="Geneious type: restriction site" misc_feature 13707..13712 /Recognition_pattern="AT^CGAT" /label="ClaI" /note="Geneious type: restriction site" misc_feature 14163..14168 /Recognition_pattern="AT^CGAT" /label="ClaI" /note="Geneious type: restriction site" misc_feature 14250..14255 /Recognition_pattern="AT^CGAT" /label="ClaI" /note="Geneious type: restriction site" misc_feature 14310..14315 /Recognition_pattern="C^YCGRG" /label="AvaI" /note="Geneious type: restriction site" misc_feature 14310..14315 /Recognition_pattern="CCC^GGG" /label="SmaI" /note="Geneious type: restriction site" misc_feature 14310..14315 /Recognition_pattern="C^CCGGG" /label="XmaI" /note="Geneious type: restriction site" misc_feature 14572..14577 /Recognition_pattern="C^YCGRG" /label="AvaI" /note="Geneious type: restriction site" ORIGIN 1 ctcgagcttc tactgggcgg ttttatggac agcaagcgaa ccggaattgc cagctggggc 61 gccctctggt aaggttggga agccctgcaa agtaaactgg atggctttct cgccgccaag 121 gatctgatgg cgcaggggat caagctctga tcaagagaca ggatgaggat cgtttcgcat 181 gattgaacaa gatggattgc acgcaggttc tccggccgct tgggtggaga ggctattcgg 241 ctatgactgg gcacaacaga caatcggctg ctctgatgcc gccgtgttcc ggctgtcagc 301 gcaggggcgc ccggttcttt ttgtcaagac cgacctgtcc ggtgccctga atgaactgca 361 agacgaggca gcgcggctat cgtggctggc cacgacgggc gttccttgcg cagctgtgct 421 cgacgttgtc actgaagcgg gaagggactg gctgctattg ggcgaagtgc cggggcagga 481 tctcctgtca tctcaccttg ctcctgccga gaaagtatcc atcatggctg atgcaatgcg 541 gcggctgcat acgcttgatc cggctacctg cccattcgac caccaagcga aacatcgcat 601 cgagcgagca cgtactcgga tggaagccgg tcttgtcgat caggatgatc tggacgaaga 661 gcatcagggg ctcgcgccag ccgaactgtt cgccaggctc aaggcgagca tgcccgacgg 721 cgaggatctc gtcgtgaccc atggcgatgc ctgcttgccg aatatcatgg tggaaaatgg 781 ccgcttttct ggattcatcg actgtggccg gctgggtgtg gcggaccgct atcaggacat 841 agcgttggct acccgtgata ttgctgaaga gcttggcggc gaatgggctg accgcttcct 901 cgtgctttac ggtatcgccg ctcccgattc gcagcgcatc gccttctatc gccttcttga 961 cgagttcttc tgaattatta acgcttacaa tttcctgatg cggtattttc tccttacgca 1021 tctgtgcggt atttcacacc gcatacaggt ggcacttttc ggggaaatgt gcgcggaacc 1081 cctatttgtt tatttttcta aatacattca aatatgtatc cgctcatgag acaataaccc 1141 tgataaatgc ttcaataata gcacgtgcta aaacttcatt tttaatttaa aaggatctag 1201 gtgaagatcc tttttgataa tctcatgacc aaaatccctt aacgtgagtt ttcgttccac 1261 tgagcgtcag accccgtaga aaagatcaaa ggatcttctt gagatccttt ttttctgcgc 1321 gtaatctgct gcttgcaaac aaaaaaacca ccgctaccag cggtggtttg tttgccggat 1381 caagagctac caactctttt tccgaaggta actggcttca gcagagcgca gataccaaat 1441 actgtccttc tagtgtagcc gtagttaggc caccacttca agaactctgt agcaccgcct 1501 acatacctcg ctctgctaat cctgttacca gtggctgctg ccagtggcga taagtcgtgt 1561 cttaccgggt tggactcaag acgatagtta ccggataagg cgcagcggtc gggctgaacg 1621 gggggttcgt gcacacagcc cagcttggag cgaacgacct acaccgaact gagataccta 1681 cagcgtgagc tatgagaaag cgccacgctt cccgaaggga gaaaggcgga caggtatccg 1741 gtaagcggca gggtcggaac aggagagcgc acgagggagc ttccaggggg aaacgcctgg 1801 tatctttata gtcctgtcgg gtttcgccac ctctgacttg agcgtcgatt tttgtgatgc 1861 tcgtcagggg ggcggagcct atggaaaaac gccagcaacg cggccttttt acggttcctg 1921 ggcttttgct ggccttttgc tcacatgttc ttgactcttc gcgatgtacg ggccagatat 1981 gtcgaccgac atgtcgcaca agtcctaagt tacgcgacag gctgccgccc tgcccttttc 2041 ctggcgtttt cttgtcgcgt gttttagtcg cataaagtag aatacttgcg actagaaccg 2101 gagacattac gccatgaaca agagcgccgc cgctggcctg ctgggctatg cccgcgtcag 2161 caccgacgac caggacttga ccaaccaacg ggccgaactg cacgcggccg gctgcaccaa 2221 gctgttttcc gagaagatca ccggcaccag gcgcgaccgc ccggagctgg ccaggatgct 2281 tgaccaccta cgccctggcg acgttgtgac agtgaccagg ctagaccgcc tggcccgcag 2341 cacccgcgac ctactggaca ttgccgagcg catccaggag gccggcgcgg gcctgcgtag 2401 cctggcagag ccgtgggccg acaccaccac gccggccggc cgcatggtgt tgaccgtgtt 2461 cgccggcatt gccgagttcg agcgttccct aatcatcgac cgcacccgga gcgggcgcga 2521 ggccgccaag gcgcgaggcg tgaagtttgg cccccgccct accctcaccc cggcacagat 2581 cgcgcacgcc cgcgagctga tcgaccagga aggccgcacc gtgaaagagg cggctgcact 2641 gcttggcgtg catcgctcga ccctgtaccg cgcacttgag cgcagcgagg aagtgacgcc 2701 caccgaggcc aggcggcgcg gtgccttccg tgaggacgca ttgaccgagg ccgacgccct 2761 ggcggccgcc gagaatgaac gccaagagga acaagcatga aaccgcacca ggacggccag 2821 gacgaaccgt ttttcattac cgaagagatc gaggcggaga tgatcgcggc cgggtacgtg 2881 ttcgagccgc ccgcgcacgt ctcaaccgtg cggctgcatg aaatcctggc cggtttgtct 2941 gatgccaagc tcgcggcctg gccggcgagc ttggccgctg aagaaaccga gcgccgccgt 3001 ctaaaaaggt gatgtgtatt tgagtaaaac agcttgcgtc atgcggtcgc tgcgtatatg 3061 atgcgatgag taaataaaca aatacgcaag gggaacgcat gaaggttatc gctgtactta 3121 accagaaagg cgggtcaggc aagacgacca tcgcaaccca tctagcccgc gccctgcaac 3181 tcgccggggc cgatgttctg ttagtcgatt ccgatcccca gggcagtgcc cgcgattggg 3241 cggccgtgcg ggaagatcaa ccgctaaccg ttgtcggcat cgaccgcccg acgattgacc 3301 gcgacgtgaa ggccatcggc cggcgcgact tcgtagtgat cgacggagcg ccccaggcgg 3361 cggacttggc tgtgtccgcg atcaaggcag ccgacttcgt gctgattccg gtgcagccaa 3421 gcccttacga catatgggcc accgccgacc tggtggagct ggttaagcag cgcattgagg 3481 tcacggatgg aaggctacaa gcggcctttg tcgtgtcgcg ggcgatcaaa ggcacgcgca 3541 tcggcggtga ggttgccgag gcgctggccg ggtacgagct gcccattctt gagtcccgta 3601 tcacgcagcg cgtgagctac ccaggcactg ccgccgccgg cacaaccgtt cttgaatcag 3661 aacccgaggg cgacgctgcc cgcgaggtcc aggcgctggc cgctgaaatt aaatcaaaac 3721 tcatttgagt taatgaggta aagagaaaat gagcaaaagc acaaacacgc taagtgccgg 3781 ccgtccgagc gcacgcagca gcaaggctgc aacgttggcc agcctggcag acacgccagc 3841 catgaagcgg gtcaactttc agttgccggc ggaggatcac accaagctga agatgtacgc 3901 ggtacgccaa ggcaagacca ttaccgagct gctatctgaa tacatcgcgc agctaccaga 3961 gtaaatgagc aaatgaataa atgagtagat gaattttagc ggctaaagga ggcggcatgg 4021 aaaatcaaga acaaccaggc accgacgccg tggaatgccc catgtgtgga ggaacgggcg 4081 gttggccagg cgtaagcggc tgggttgtct gccggccctg caatggcact ggaaccccca 4141 agcccgagga atcggcgtga gcggtcgcaa accatccggc ccggtacaaa tcggcgcggc 4201 gctgggtgat gacctggtgg agaagttgaa ggcggcgcag gccgcccagc ggcaacgcat 4261 cgaggcagaa gcacgccccg gtgaatcgtg gcaagcggcc gctgatcgaa tccgcaaaga 4321 atcccggcaa ccgccggcag ccggtgcgcc gtcgattagg aagccgccca agggcgacga 4381 gcaaccagat tttttcgttc cgatgctcta tgacgtgggc acccgcgata gtcgcagcat 4441 catggacgtg gccgttttcc gtctgtcgaa gcgtgaccga cgagctggcg aggtgatccg 4501 ctacgagctt ccagacgggc acgtagaggt ttccgcaggg ccggccggca tggcgagtgt 4561 gtgggattac gacctggtac tgatggcggt ttcccatcta accgaatcca tgaaccgata 4621 ccgggaaggg aagggagaca agcccggccg cgtgttccgt ccacacgttg cggacgtact 4681 caagttctgc cggcgagccg atggcggaaa gcagaaagac gacctggtag aaacctgcat 4741 tcggttaaac accacgcacg ttgccatgca gcgtacgaag aaggccaaga acggccgcct 4801 ggtgacggta tccgagggtg aagccttgat tagccgctac aagatcgtaa agagcgaaac 4861 cgggcggccg gagtacatcg agatcgagtt agctgattgg atgtaccgcg agatcacaga 4921 aggcaagaac ccggacgtgc tgacggttca ccccgattac tttttgatcg atcccggcat 4981 cggccgtttt ctctaccgcc tggcacgccg cgccgcaggc aaggcagaag ccagatggtt 5041 gttcaagacg atctacgaac gcagtggcag cgccggagag ttcaagaagt tctgtttcac 5101 cgtgcgcaag ctgatcgggt caaatgacct gccggagtac gatttgaagg aggaggcggg 5161 gcaggctggc ccgatcctag tcatgcgcta ccgcaacctg atcgagggcg aagcatccgc 5221 cggttcctaa tgtacggagc agatgctagg gcaaattgcc ctagcagggg aaaaaggtcg 5281 aaaaggtctc tttcctgtgg atagcacgta cattgggaac ccaaagccgt acattgggaa 5341 ccggaacccg tacattggga acccaaagcc gtacattggg aaccggtcac acatgtaagt 5401 gactgatata aaagagaaaa aaggcgattt ttccgcctaa aactctttaa aacttattaa 5461 aactcttaaa acccgcctgg cctgtgcata actgtctggc cagcgcacag ccgaagagct 5521 gcaaaaagcg cctacccttc ggtcgctgcg ctccctacgc cccgccgctt cgcgtcggcc 5581 tatcgcggcc gctggccgct caaaaatggc tggcctacgg ccaggcaatc taccagggcg 5641 cggacaagcc gcgccgtcgc cactcgaccg ccggcgccca catcaaggca cctctagatg 5701 gcaggatata ttgtggtgta aacagtttaa acacatagat gacaccgcgc gcgataattt 5761 atcctagttt gcgcgctata ttttgttttc tatcgcgtat taaatgtata attgcgggac 5821 tctaatcata aaaacccatc tcataaataa cgtcatgcat tacatgttaa ttattacatg 5881 cttaacgtaa ttcaacagaa attatatgat aatcatcgca agaccggcaa caggattcaa 5941 tcttaagaaa ctttattgcc aaatgtttga acgatcgggg aaattcgagc tcggtagcaa 6001 ttcccgaggc tgtagccgac gatggtgcgc caggagagtt gttgatttat cactgctttc 6061 cgccttgctg aggtttctca ccgaagttca ttcctgtcca tctcttttgg agaagaaaag 6121 cggcagattt tggcttccta tctctagtga agatgccttt cttatttcca cccaccctca 6181 gaatgccctg tgaggttgca aagtcagcga agttccaaac ttgttcaccg accacggcgg 6241 aaactctgtc aaagaccctg tggtacatat ccaaccatgc gcactggtac tcttcgctcc 6301 acatatctgt atacatagaa tggagtccag caagagtgtc cacgccatac tcggtgataa 6361 tgatcggttg gtgcagcttt tcctgccagg cgagcaactc tttctcaaga actttttctg 6421 ctgtctccag atcgcctgat tggacgtacc agccatagta tctattcaaa cagagcacat 6481 cgaacaggtc ggaaatagta tctgtatgag cgtcgcaaaa cataacgttg acacaggtga 6541 taggtctagt tggatcaagt tttctggtgg cttcagcgag cggtgcgaaa tactccctgg 6601 ctccctgagg tcttgtgtct ggttcattag caatgctcca catcacaaca gacgggtggt 6661 tcttatccct tgcgatcaac tccttaatag cttgaagatg ggcctgttga gtctcaccat 6721 tcacagcttc ctctgagtac agttctttag gcttgtttcc tgcctcgaag ccgataccga 6781 gggaaagatt aaatccaacg gcagcggtct cgtcaatgac cacgatgccg tgttcatctg 6841 cccaatccaa catctcttcg gcataagggt agtgagatgt tctgtagctg ttagcaccaa 6901 tccagtccat gagggcatga tcgtgaacca tcaggacatt gtcgaatcct ttgcccctca 6961 aatctgcgtc ctcatgtcta ccaaatccag tgaaataaaa tggcttgtgg ttgataagga 7021 actgttcgcc cttcacagca actgatctaa taccgactct gagcgggtag atatcgcact 7081 cggtctgtga tttggctgtc acacaaagct catacaggta tccttcgcct ggttgccaca 7141 agtgagggtt aaccacttgg agagtaccgg aggttccttg gcctgtagca acgacttgtt 7201 gatctgcgtc cctaagctcc acagaaacat caccattagc gacaacctgc acatcaacaa 7261 attttggtca tatattagaa aagttataaa ttaaaatata cacacttata aactacagaa 7321 aagcaattgc tatatactac attcttttat tttgaaaaaa atatttgaaa tattatatta 7381 ctactaatta atgataatta ttatatatat atcaaaggta gaagcagaaa cttacctgcc 7441 agtcaacgct ggcatggttg caatcttgtg cgacgtgagt cacaacggta atgtcatcga 7501 cccatgtatt cggagtggta tacagcatca ctgatctatg gattccagcg tagttaaaga 7561 agtcgtggaa ataggactgt ttcttcttac cattttcatc tgtaataacc atgcctggcg 7621 ggatagtttg ccagttcaac tcattgttca cacaaacggt aatcctgaca gactttccgg 7681 cgatcacgta aggtgtaaca tcagcttcaa atggagtata gccaccctga tgctccatga 7741 cttcttgatt gttcacccaa actttgccgt agtgggtgac tgcgtcgaat ctgagaacaa 7801 tcctctgtcc ggcccaaccc ttagggataa acacttctct ttgataccag acatttccag 7861 cgtagttcct aatatctgca tcggcgaact ggtcattaaa gctgcccggc acagcgattg 7921 ctcttgactc ttgaagggcg gattcccacc atctctgatc aattccgcag ttctctctgt 7981 ccagagagaa agcccacaag ccatcgagtt tcttgatttc ccttgttgga gtctcaacag 8041 gtctaagcat tgttggtacc cagcttgggt ctagtcgtat taagagatag atttgtagag 8101 agagactggt gatttcagcg tgtcctctcc aaatgaaatg aacttcctta tatagaggaa 8161 ggtcttgcga aggatagtgg gattgtgcgt catcccttac gtcagtggag atatcacatc 8221 aatccacttg ctttgaagac gtggttggaa cgtcttcttt ttccacgatg ctcctcgtgg 8281 gtgggggtcc atctttggga ccactgtcgg cagaggcatc ttgaacgata gcctttcctt 8341 tatcgcaatg atggcatttg taggtgccac cttccttttc tactgtcctt ttgatgaagt 8401 gacagatagc tgggcaatgg aatccgagga ggtttcccga tattaccctt tgttgaaaag 8461 tctcaatagc cctttggtct tctgagactg tatctttgat attcttggag tagacgagag 8521 tgtcgtgctc caccatgtta tcacatcaat ccacttgctt tgaagacgtg gttggaacgt 8581 cttctttttc cacgatgctc ctcgtgggtg ggggtccatc tttgggacca ctgtcggcag 8641 aggcatcttg aacgatagcc tttcctttat cgcaatgatg gcatttgtag gtgccacctt 8701 ccttttctac tgtccttttg atgaagtgac agatagctgg gcaatggaat ccgaggaggt 8761 ttcccgatat taccctttgt tgaaaagtct caatagccct ttggtcttct gagactgtat 8821 ctttgatatt cttggagtag acgagagtgt cgtgctccac cattacatag gcccatcgga 8881 gctaacgcag tgaattcaga aatctcaaaa ttccggcaga acaattttga atctcgatcc 8941 gtagaaacga gacggtcatt gttttagttc caccacgatt atatttgaaa tttacgtgag 9001 tgtgagtgag acttgcataa gaaaataaaa tctttagttg ggaaaaaatt caataatata 9061 aatgggcttg agaaggaagc gagggatagg cctttttcta aaataggccc atttaagcta 9121 ttaacaatct tcaaaagtac cacagcgctt aggtaaagaa agcagctgag tttatatatg 9181 gttagagacg aagtagtgat tggatggcag gtggaagaat ggacacctgc gagagtttta 9241 gagctagaaa tagcaagtta aaataaggct agtccgttat caacttgaaa aagtggcacc 9301 gagtcggtgc tttttttaaa gcttactgcg ttagctccga tgggcctatg taatggtgga 9361 gcacgacact ctcgtctact ccaagaatat caaagataca gtctcagaag accaaagggc 9421 tattgagact tttcaacaaa gggtaatatc gggaaacctc ctcggattcc attgcccagc 9481 tatctgtcac ttcatcaaaa ggacagtaga aaaggaaggt ggcacctaca aatgccatca 9541 ttgcgataaa ggaaaggcta tcgttcaaga tgcctctgcc gacagtggtc ccaaagatgg 9601 acccccaccc acgaggagca tcgtggaaaa agaagacgtt ccaaccacgt cttcaaagca 9661 agtggattga tgtgataaca tggtggagca cgacactctc gtctactcca agaatatcaa 9721 agatacagtc tcagaagacc aaagggctat tgagactttt caacaaaggg taatatcggg 9781 aaacctcctc ggattccatt gcccagctat ctgtcacttc atcaaaagga cagtagaaaa 9841 ggaaggtggc acctacaaat gccatcattg cgataaagga aaggctatcg ttcaagatgc 9901 ctctgccgac agtggtccca aagatggacc cccacccacg aggagcatcg tggaaaaaga 9961 agacgttcca accacgtctt caaagcaagt ggattgatgt gatatctcca ctgacgtaag 10021 ggatgacgca caatcccact atccttcgca agaccttcct ctatataagg aagttcattt 10081 catttggaga ggacacgctg aaatcaccag tctctctcta caaatctatc tcttaatacg 10141 actcactata gggagaccca agctggctag caacaatgga taagaagtac tctatcggac 10201 tcgatatcgg aactaactct gttggatggg ctgtgatcac cgatgagtac aaggtgccat 10261 ctaagaagtt caaggttctc ggaaacaccg ataggcactc tatcaagaaa aaccttatcg 10321 gtgctctcct cttcgattct ggtgaaactg ctgaggctac cagactcaag agaaccgcta 10381 gaagaaggta caccagaaga aagaacagga tctgctacct ccaagagatt ttctctaacg 10441 agatggctaa agtggatgat tcattcttcc acaggctcga agagtcattc ctcgtggaag 10501 aagataagaa gcacgagagg caccctatct tcggaaacat cgttgatgag gtggcatacc 10561 acgagaagta ccctactatc taccacctca gaaagaagct cgttgattct actgataagg 10621 ctgatctcag gctcatctac ctcgctctcg ctcacatgat caagttcaga ggacacttcc 10681 tcatcgaggg tgatctcaac cctgataact ctgatgtgga taagttgttc atccagctcg 10741 tgcagaccta caaccagctt ttcgaagaga accctatcaa cgcttcaggt gtggatgcta 10801 aggctatcct ctctgctagg ctctctaagt caagaaggct tgagaacctc attgctcagc 10861 tccctggtga gaagaagaac ggacttttcg gaaacttgat cgctctctct ctcggactca 10921 cccctaactt caagtctaac ttcgatctcg ctgaggatgc aaagctccag ctctcaaagg 10981 atacctacga tgatgatctc gataacctcc tcgctcagat cggagatcag tacgctgatt 11041 tgttcctcgc tgctaagaac ctctctgatg ctatcctcct cagtgatatc ctcagggtga 11101 acaccgagat caccaaggct ccactttctg cttctatgat caagagatac gatgagcacc 11161 accaggatct cacacttctc aaggctcttg ttagacagca gctcccagag aagtacaaag 11221 aaatcttctt cgatcagtct aagaacggat acgctggtta catcgatggt ggtgcatctc 11281 aagaagagtt ctacaagttc atcaagccaa tcttggagaa gatggatgga accgaggaac 11341 tcctcgtgaa gctcaataga gaggatctcc ttaggaagca gaggaccttc gataacggat 11401 ctatccctca tcagatccac ctcggagagt tgcacgctat ccttagaagg caagaggatt 11461 tctacccatt cctcaaggat aacagagaga agattgagaa gatcctcacc ttcagaatcc 11521 cttactacgt gggacctctc gctagaggaa actcaagatt cgcttggatg accagaaagt 11581 ctgaggaaac catcacccct tggaacttcg aagaggtggt ggataagggt gctagtgctc 11641 agtctttcat cgagaggatg accaacttcg ataagaacct tcctaacgag aaggtgctcc 11701 ctaagcactc tttgctctac gagtacttca ccgtgtacaa cgagttgacc aaggttaagt 11761 acgtgaccga gggaatgagg aagcctgctt ttttgtcagg tgagcaaaag aaggctatcg 11821 ttgatctctt gttcaagacc aacagaaagg tgaccgtgaa gcagctcaaa gaggattact 11881 tcaagaaaat cgagtgcttc gattcagtgg aaatctctgg tgttgaggat aggttcaacg 11941 catctctcgg aacctaccac gatctcctca agatcattaa ggataaggat ttcttggata 12001 acgaggaaaa cgaggatatc ttggaggata tcgttcttac cctcaccctc ttcgaggata 12061 gagagatgat agaagaaagg ctcaagacct acgctcatct cttcgatgat aaggtgatga 12121 agcagttgaa gagaagaaga tacactggtt ggggaaggct ctcaagaaag ctcattaacg 12181 gaatcaggga taagcagtct ggaaagacaa tccttgattt cctcaagtct gatggattcg 12241 ctaacagaaa cttcatgcag ctcatccacg atgattctct cacctttaaa gaggatatcc 12301 agaaggctca ggtttcagga cagggtgata gtctccatga gcatatcgct aacctcgctg 12361 gatcccctgc aatcaagaag ggaatcctcc agactgtgaa gattgtggat gagttggtga 12421 aggtgatggg acacaagcct gagaacatcg tgatcgaaat ggctagagag aaccagacca 12481 ctcagaaggg acagaagaac tctagggaaa ggatgaagag gatcgaggaa ggtatcaaag 12541 agcttggatc tcagatcctc aaagagcacc ctgttgagaa cactcagctc cagaacgaga 12601 agctctacct ctactacttg cagaacggaa gggatatgta tgtggatcaa gagcttgata 12661 ttaacaggct ctctgattac gatgttgatc atatcgtgcc acagtctttt atcaaagatg 12721 attctatcga taacaaggtg ctcactaggt ctgataagaa caggggtaag agtgataacg 12781 tgccaagtga agaggttgtg aagaaaatga agaactattg gaggcagctc ctcaacgcta 12841 agctcatcac tcagagaaag ttcgataact tgaccaaggc tgagagggga ggactctctg 12901 aattggataa ggcaggattc atcaagagac agctcgtgga aaccaggcag atcaccaaac 12961 atgtggcaca gatcctcgat tctaggatga acaccaagta cgatgagaac gataagttga 13021 tcagggaagt gaaggttatc accctcaagt caaagctcgt gtctgatttc agaaaggatt 13081 tccaattcta caaggtgagg gaaatcaaca actaccacca cgctcacgat gcttacctta 13141 acgctgttgt tggaaccgct ctcatcaaga agtatccaaa gttggagtct gagttcgtgt 13201 acggtgatta taaggtgtac gatgtgagga agatgatcgc taagtctgag caagagatcg 13261 gaaaggctac cgctaagtat ttcttctact ctaacatcat gaatttcttc aagaccgaga 13321 tcactctcgc taacggtgag atcagaaaga ggccactcat cgagacaaac ggtgaaacag 13381 gtgagatcgt gtgggataag ggaagggatt tcgctaccgt tagaaaggtg ctctctatgc 13441 ctcaggtgaa catcgttaag aaaaccgagg tgcagaccgg tggattctct aaagagtcta 13501 tcctccctaa gaggaactct gataagctca ttgctaggaa gaaggattgg gaccctaaga 13561 aatacggtgg tttcgattct cctaccgtgg cttactctgt tctcgttgtg gctaaggttg 13621 agaagggaaa gagtaagaag ctcaagtctg ttaaggaact tctcggaatc actatcatgg 13681 aaaggtcatc tttcgagaag aacccaatcg atttccttga ggctaaggga tacaaagagg 13741 ttaagaagga tctcatcatc aagctcccaa agtactcact tttcgagttg gagaacggta 13801 gaaagaggat gctcgcttct gctggtgagc ttcaaaaggg aaacgagctt gctctcccat 13861 ctaagtacgt taactttctt tacctcgctt ctcactacga gaagttgaag ggatctccag 13921 aagataacga gcagaagcaa cttttcgttg agcagcacaa gcactacttg gatgagatca 13981 tcgagcagat cagtgagttc tctaaaaggg tgatcctcgc tgatgcaaac ctcgataagg 14041 tgttgtctgc ttacaacaag cacagagata agcctatcag ggaacaggca gagaacatca 14101 tccatctctt cacccttacc aacctcggtg ctcctgctgc tttcaagtac ttcgatacaa 14161 ccatcgatag gaagagatac acctctacca aagaagtgct cgatgctacc ctcatccatc 14221 agtctatcac tggactctac gagactagga tcgatctctc acagcttgga ggtgatccta 14281 agaagaaaag aaaggttaga tcttgatgac ccgggtctcc ataataatgt gtgagtagtt 14341 cccagataag ggaattaggg ttcctatagg gtttcgctca tgtgttgagc atataagaaa 14401 cccttagtat gtatttgtat ttgtaaaata cttctatcaa taaaatttct aattcctaaa 14461 accaaaatcc agtactaaaa tccagatccc ccgaattaag gccttgacag gatatattgg 14521 cgggtaaacc taagagaaaa gagcgtttat tagaataacg gatatttaaa actcgag //