LOCUS p53 13562 bp DNA circular 22-AUG-2017 DEFINITION . ACCESSION urn.local...8a-7o8q4x2 KEYWORDS . SOURCE ORGANISM . FEATURES Location/Qualifiers misc_feature 179..973 /label="Kan R" primer 426..445 /label="TDNA-pUC Rev primer" misc_feature 1273..1946 /label="pUC Ori" primer 1827..1846 /label="pUC-pVS Forw4 primer" misc_feature 1981..1986 /Recognition_pattern="G^TCGAC" /label="SalI" /note="Geneious type: restriction site" insertion 1987..5692 /label="pVS Ori" primer 2348..2367 /label="pUC-pVS Rev4 primer" primer 5481..5500 /label="pVS-TDNA Forw primer" misc_feature 5693..5698 /Recognition_pattern="T^CTAGA" /label="XbaI" /note="Geneious type: restriction site" misc_feature 5699..5724 /label="T-DNA LB" misc_feature complement(5725..6000) /label="NOS terminator" misc_feature complement(6001..7033) /label="Hygromycin R" primer 6039..6058 /label="pVS-TDNA Rev primer" misc_feature complement(7040..7870) /label="35 S promoter" primer 7288..7307 /label="TDNA-pUC Forw primer" misc_feature 7871..7876 /Recognition_pattern="G^AATTC" /label="EcoRI" /note="Geneious type: restriction site" allele 7877..8181 /label="AtU6" misc_feature 8067..8072 /Recognition_pattern="AGG^CCT" /label="StuI" /note="Geneious type: restriction site" misc_feature 8186..8192 /label="AarI" misc_feature 8203..8209 /label="AarI" misc_feature 8297..8302 /label="Barcode" misc_feature 8304..8309 /Recognition_pattern="A^AGCTT" /label="HindIII" /note="Geneious type: restriction site" misc_feature 8338..9118 /label="35S" misc_feature 9119..9137 /label="T7" misc_feature 9151..9156 /Recognition_pattern="G^CTAGC" /label="NheI" /note="Geneious type: restriction site" C_region 9157..13294 /label="AtCas9" misc_feature 11345..11350 /Recognition_pattern="G^GATCC" /label="BamHI" /note="Geneious type: restriction site" primer 13193..13215 /label="ATSF8 primer" misc_feature 13283..13288 /Recognition_pattern="A^GATCT" /label="BglII" /note="Geneious type: restriction site" misc_feature 13295..13300 /Recognition_pattern="C^CCGGG" /label="XmaI" /note="Geneious type: restriction site" misc_feature 13484..13489 /Recognition_pattern="AGG^CCT" /label="StuI" /note="Geneious type: restriction site" misc_feature 13490..13513 /label="T-DNA RB" misc_feature 13557..13562 /label="XhoI" ORIGIN 1 ctcgagcttc tactgggcgg ttttatggac agcaagcgaa ccggaattgc cagctggggc 61 gccctctggt aaggttggga agccctgcaa agtaaactgg atggctttct cgccgccaag 121 gatctgatgg cgcaggggat caagctctga tcaagagaca ggatgaggat cgtttcgcat 181 gattgaacaa gatggattgc acgcaggttc tccggccgct tgggtggaga ggctattcgg 241 ctatgactgg gcacaacaga caatcggctg ctctgatgcc gccgtgttcc ggctgtcagc 301 gcaggggcgc ccggttcttt ttgtcaagac cgacctgtcc ggtgccctga atgaactgca 361 agacgaggca gcgcggctat cgtggctggc cacgacgggc gttccttgcg cagctgtgct 421 cgacgttgtc actgaagcgg gaagggactg gctgctattg ggcgaagtgc cggggcagga 481 tctcctgtca tctcaccttg ctcctgccga gaaagtatcc atcatggctg atgcaatgcg 541 gcggctgcat acgcttgatc cggctacctg cccattcgac caccaagcga aacatcgcat 601 cgagcgagca cgtactcgga tggaagccgg tcttgtcgat caggatgatc tggacgaaga 661 gcatcagggg ctcgcgccag ccgaactgtt cgccaggctc aaggcgagca tgcccgacgg 721 cgaggatctc gtcgtgaccc atggcgatgc ctgcttgccg aatatcatgg tggaaaatgg 781 ccgcttttct ggattcatcg actgtggccg gctgggtgtg gcggaccgct atcaggacat 841 agcgttggct acccgtgata ttgctgaaga gcttggcggc gaatgggctg accgcttcct 901 cgtgctttac ggtatcgccg ctcccgattc gcagcgcatc gccttctatc gccttcttga 961 cgagttcttc tgaattatta acgcttacaa tttcctgatg cggtattttc tccttacgca 1021 tctgtgcggt atttcacacc gcatacaggt ggcacttttc ggggaaatgt gcgcggaacc 1081 cctatttgtt tatttttcta aatacattca aatatgtatc cgctcatgag acaataaccc 1141 tgataaatgc ttcaataata gcacgtgcta aaacttcatt tttaatttaa aaggatctag 1201 gtgaagatcc tttttgataa tctcatgacc aaaatccctt aacgtgagtt ttcgttccac 1261 tgagcgtcag accccgtaga aaagatcaaa ggatcttctt gagatccttt ttttctgcgc 1321 gtaatctgct gcttgcaaac aaaaaaacca ccgctaccag cggtggtttg tttgccggat 1381 caagagctac caactctttt tccgaaggta actggcttca gcagagcgca gataccaaat 1441 actgtccttc tagtgtagcc gtagttaggc caccacttca agaactctgt agcaccgcct 1501 acatacctcg ctctgctaat cctgttacca gtggctgctg ccagtggcga taagtcgtgt 1561 cttaccgggt tggactcaag acgatagtta ccggataagg cgcagcggtc gggctgaacg 1621 gggggttcgt gcacacagcc cagcttggag cgaacgacct acaccgaact gagataccta 1681 cagcgtgagc tatgagaaag cgccacgctt cccgaaggga gaaaggcgga caggtatccg 1741 gtaagcggca gggtcggaac aggagagcgc acgagggagc ttccaggggg aaacgcctgg 1801 tatctttata gtcctgtcgg gtttcgccac ctctgacttg agcgtcgatt tttgtgatgc 1861 tcgtcagggg ggcggagcct atggaaaaac gccagcaacg cggccttttt acggttcctg 1921 ggcttttgct ggccttttgc tcacatgttc ttgactcttc gcgatgtacg ggccagatat 1981 gtcgaccgac atgtcgcaca agtcctaagt tacgcgacag gctgccgccc tgcccttttc 2041 ctggcgtttt cttgtcgcgt gttttagtcg cataaagtag aatacttgcg actagaaccg 2101 gagacattac gccatgaaca agagcgccgc cgctggcctg ctgggctatg cccgcgtcag 2161 caccgacgac caggacttga ccaaccaacg ggccgaactg cacgcggccg gctgcaccaa 2221 gctgttttcc gagaagatca ccggcaccag gcgcgaccgc ccggagctgg ccaggatgct 2281 tgaccaccta cgccctggcg acgttgtgac agtgaccagg ctagaccgcc tggcccgcag 2341 cacccgcgac ctactggaca ttgccgagcg catccaggag gccggcgcgg gcctgcgtag 2401 cctggcagag ccgtgggccg acaccaccac gccggccggc cgcatggtgt tgaccgtgtt 2461 cgccggcatt gccgagttcg agcgttccct aatcatcgac cgcacccgga gcgggcgcga 2521 ggccgccaag gcgcgaggcg tgaagtttgg cccccgccct accctcaccc cggcacagat 2581 cgcgcacgcc cgcgagctga tcgaccagga aggccgcacc gtgaaagagg cggctgcact 2641 gcttggcgtg catcgctcga ccctgtaccg cgcacttgag cgcagcgagg aagtgacgcc 2701 caccgaggcc aggcggcgcg gtgccttccg tgaggacgca ttgaccgagg ccgacgccct 2761 ggcggccgcc gagaatgaac gccaagagga acaagcatga aaccgcacca ggacggccag 2821 gacgaaccgt ttttcattac cgaagagatc gaggcggaga tgatcgcggc cgggtacgtg 2881 ttcgagccgc ccgcgcacgt ctcaaccgtg cggctgcatg aaatcctggc cggtttgtct 2941 gatgccaagc tcgcggcctg gccggcgagc ttggccgctg aagaaaccga gcgccgccgt 3001 ctaaaaaggt gatgtgtatt tgagtaaaac agcttgcgtc atgcggtcgc tgcgtatatg 3061 atgcgatgag taaataaaca aatacgcaag gggaacgcat gaaggttatc gctgtactta 3121 accagaaagg cgggtcaggc aagacgacca tcgcaaccca tctagcccgc gccctgcaac 3181 tcgccggggc cgatgttctg ttagtcgatt ccgatcccca gggcagtgcc cgcgattggg 3241 cggccgtgcg ggaagatcaa ccgctaaccg ttgtcggcat cgaccgcccg acgattgacc 3301 gcgacgtgaa ggccatcggc cggcgcgact tcgtagtgat cgacggagcg ccccaggcgg 3361 cggacttggc tgtgtccgcg atcaaggcag ccgacttcgt gctgattccg gtgcagccaa 3421 gcccttacga catatgggcc accgccgacc tggtggagct ggttaagcag cgcattgagg 3481 tcacggatgg aaggctacaa gcggcctttg tcgtgtcgcg ggcgatcaaa ggcacgcgca 3541 tcggcggtga ggttgccgag gcgctggccg ggtacgagct gcccattctt gagtcccgta 3601 tcacgcagcg cgtgagctac ccaggcactg ccgccgccgg cacaaccgtt cttgaatcag 3661 aacccgaggg cgacgctgcc cgcgaggtcc aggcgctggc cgctgaaatt aaatcaaaac 3721 tcatttgagt taatgaggta aagagaaaat gagcaaaagc acaaacacgc taagtgccgg 3781 ccgtccgagc gcacgcagca gcaaggctgc aacgttggcc agcctggcag acacgccagc 3841 catgaagcgg gtcaactttc agttgccggc ggaggatcac accaagctga agatgtacgc 3901 ggtacgccaa ggcaagacca ttaccgagct gctatctgaa tacatcgcgc agctaccaga 3961 gtaaatgagc aaatgaataa atgagtagat gaattttagc ggctaaagga ggcggcatgg 4021 aaaatcaaga acaaccaggc accgacgccg tggaatgccc catgtgtgga ggaacgggcg 4081 gttggccagg cgtaagcggc tgggttgtct gccggccctg caatggcact ggaaccccca 4141 agcccgagga atcggcgtga gcggtcgcaa accatccggc ccggtacaaa tcggcgcggc 4201 gctgggtgat gacctggtgg agaagttgaa ggcggcgcag gccgcccagc ggcaacgcat 4261 cgaggcagaa gcacgccccg gtgaatcgtg gcaagcggcc gctgatcgaa tccgcaaaga 4321 atcccggcaa ccgccggcag ccggtgcgcc gtcgattagg aagccgccca agggcgacga 4381 gcaaccagat tttttcgttc cgatgctcta tgacgtgggc acccgcgata gtcgcagcat 4441 catggacgtg gccgttttcc gtctgtcgaa gcgtgaccga cgagctggcg aggtgatccg 4501 ctacgagctt ccagacgggc acgtagaggt ttccgcaggg ccggccggca tggcgagtgt 4561 gtgggattac gacctggtac tgatggcggt ttcccatcta accgaatcca tgaaccgata 4621 ccgggaaggg aagggagaca agcccggccg cgtgttccgt ccacacgttg cggacgtact 4681 caagttctgc cggcgagccg atggcggaaa gcagaaagac gacctggtag aaacctgcat 4741 tcggttaaac accacgcacg ttgccatgca gcgtacgaag aaggccaaga acggccgcct 4801 ggtgacggta tccgagggtg aagccttgat tagccgctac aagatcgtaa agagcgaaac 4861 cgggcggccg gagtacatcg agatcgagtt agctgattgg atgtaccgcg agatcacaga 4921 aggcaagaac ccggacgtgc tgacggttca ccccgattac tttttgatcg atcccggcat 4981 cggccgtttt ctctaccgcc tggcacgccg cgccgcaggc aaggcagaag ccagatggtt 5041 gttcaagacg atctacgaac gcagtggcag cgccggagag ttcaagaagt tctgtttcac 5101 cgtgcgcaag ctgatcgggt caaatgacct gccggagtac gatttgaagg aggaggcggg 5161 gcaggctggc ccgatcctag tcatgcgcta ccgcaacctg atcgagggcg aagcatccgc 5221 cggttcctaa tgtacggagc agatgctagg gcaaattgcc ctagcagggg aaaaaggtcg 5281 aaaaggtctc tttcctgtgg atagcacgta cattgggaac ccaaagccgt acattgggaa 5341 ccggaacccg tacattggga acccaaagcc gtacattggg aaccggtcac acatgtaagt 5401 gactgatata aaagagaaaa aaggcgattt ttccgcctaa aactctttaa aacttattaa 5461 aactcttaaa acccgcctgg cctgtgcata actgtctggc cagcgcacag ccgaagagct 5521 gcaaaaagcg cctacccttc ggtcgctgcg ctccctacgc cccgccgctt cgcgtcggcc 5581 tatcgcggcc gctggccgct caaaaatggc tggcctacgg ccaggcaatc taccagggcg 5641 cggacaagcc gcgccgtcgc cactcgaccg ccggcgccca catcaaggca cctctagatg 5701 gcaggatata ttgtggtgta aacagtttaa acccgatcta gtaacataga tagcaccgcg 5761 cgcgataatt tatcctagtt tgcgcgctat attttgtttt ctatcgcgta ttaaatgtat 5821 aattgcggga ctctaatcat aaaaacccat ctcataaata acgtcatgca ttacatgtta 5881 attattacat gcttaacgta attcaacaga aattatatga taatcatcgc aagaccggca 5941 acaggattca atcttaagaa actttattgc caaatgtttg aacgatcggg gagtttaaac 6001 ttactattcc tttgccctcg gacgagtgct ggggcgtcgg tttccactat cggcgagtac 6061 ttctacacag ccatcggtcc agacggccgc gcttctgcgg gcgatttgtg tacgcccgac 6121 agtcccggct ccggatcgga cgattgcgtc gcatcgaccc tgcgcccaag ctgcatcatc 6181 gaaattgccg tcaaccaagc tctgatagag ttggtcaaga ccaatgcgga gcatatacgc 6241 ccggagccgc ggcgatcctg caagctccgg atgcctccgc tcgaagtagc gcgtctgctg 6301 ctccatacaa gccaaccacg gcctccagaa gaagatgttg gcgacctcgt attgggaatc 6361 cccgaacatc gcctcgctcc agtcaatgac cgctgttatg cggccattgt ccgtcaggac 6421 attgttggag ccgaaatccg cgtgcacgag gtgccggact tcggggcagt cctcggccca 6481 aagcatcagc tcatcgagag cctgcgcgac ggacgcactg acggtgtcgt ccatcacagt 6541 ttgccagtga tacacatggg gatcagcaat cgcgcatatg aaatcacgcc atgtagtgta 6601 ttgaccgatt ccttgcggtc cgaatgggcc gaacccgctc gtctggctaa gatcggccgc 6661 agcgatcgca tccatggcct ccgcgaccgg ctgcagaaca gcgggcagtt cggtttcagg 6721 caggtcttgc aacgtgacac cctgtgcacg gcgggagatg caataggtca ggctctcgct 6781 gaactcccca atgtcaagca cttccggaat cgggagcgcg gccgatgcaa agtgccgata 6841 aacataacga tctttgtaga aaccatcggc gcagctattt acccgcagga catatccacg 6901 ccctcctaca tcgaagctga aagcacgaga ttcttcgccc tccgagagct gcatcaggtc 6961 ggagacgctg tcgaactttt cgatcagaaa cttctcgaca gacgtcgcgg tgagttcagg 7021 ctttttcatt gttggtaccc agcttgggtc tagtcgtatt aagagataga tttgtagaga 7081 gagactggtg atttcagcgt gtcctctcca aatgaaatga acttccttat atagaggaag 7141 gtcttgcgaa ggatagtggg attgtgcgtc atcccttacg tcagtggaga tatcacatca 7201 atccacttgc tttgaagacg tggttggaac gtcttctttt tccacgatgc tcctcgtggg 7261 tgggggtcca tctttgggac cactgtcggc agaggcatct tgaacgatag cctttccttt 7321 atcgcaatga tggcatttgt aggtgccacc ttccttttct actgtccttt tgatgaagtg 7381 acagatagct gggcaatgga atccgaggag gtttcccgat attacccttt gttgaaaagt 7441 ctcaatagcc ctttggtctt ctgagactgt atctttgata ttcttggagt agacgagagt 7501 gtcgtgctcc accatgttat cacatcaatc cacttgcttt gaagacgtgg ttggaacgtc 7561 ttctttttcc acgatgctcc tcgtgggtgg gggtccatct ttgggaccac tgtcggcaga 7621 ggcatcttga acgatagcct ttcctttatc gcaatgatgg catttgtagg tgccaccttc 7681 cttttctact gtccttttga tgaagtgaca gatagctggg caatggaatc cgaggaggtt 7741 tcccgatatt accctttgtt gaaaagtctc aatagccctt tggtcttctg agactgtatc 7801 tttgatattc ttggagtaga cgagagtgtc gtgctccacc attacatagg cccatcggag 7861 ctaacgcagt gaattcagaa atctcaaaat tccggcagaa caattttgaa tctcgatccg 7921 tagaaacgag acggtcattg ttttagttcc accacgatta tatttgaaat ttacgtgagt 7981 gtgagtgaga cttgcataag aaaataaaat ctttagttgg gaaaaaattc aataatataa 8041 atgggcttga gaaggaagcg agggataggc ctttttctaa aataggccca tttaagctat 8101 taacaatctt caaaagtacc acagcgctta ggtaaagaaa gcagctgagt ttatatatgg 8161 ttagagacga agtagtgatt ggatggcagg tggaagaatg gacacctgcg agagttttag 8221 agctagaaat agcaagttaa aataaggcta gtccgttatc aacttgaaaa agtggcaccg 8281 agtcggtgct ttttttcgat gtaaagctta ctgcgttagc tccgatgggc ctatgtaatg 8341 gtggagcacg acactctcgt ctactccaag aatatcaaag atacagtctc agaagaccaa 8401 agggctattg agacttttca acaaagggta atatcgggaa acctcctcgg attccattgc 8461 ccagctatct gtcacttcat caaaaggaca gtagaaaagg aaggtggcac ctacaaatgc 8521 catcattgcg ataaaggaaa ggctatcgtt caagatgcct ctgccgacag tggtcccaaa 8581 gatggacccc cacccacgag gagcatcgtg gaaaaagaag acgttccaac cacgtcttca 8641 aagcaagtgg attgatgtga taacatggtg gagcacgaca ctctcgtcta ctccaagaat 8701 atcaaagata cagtctcaga agaccaaagg gctattgaga cttttcaaca aagggtaata 8761 tcgggaaacc tcctcggatt ccattgccca gctatctgtc acttcatcaa aaggacagta 8821 gaaaaggaag gtggcaccta caaatgccat cattgcgata aaggaaaggc tatcgttcaa 8881 gatgcctctg ccgacagtgg tcccaaagat ggacccccac ccacgaggag catcgtggaa 8941 aaagaagacg ttccaaccac gtcttcaaag caagtggatt gatgtgatat ctccactgac 9001 gtaagggatg acgcacaatc ccactatcct tcgcaagacc ttcctctata taaggaagtt 9061 catttcattt ggagaggaca cgctgaaatc accagtctct ctctacaaat ctatctctta 9121 atacgactca ctatagggag acccaagctg gctagcaaca atggataaga agtactctat 9181 cggactcgat atcggaacta actctgttgg atgggctgtg atcaccgatg agtacaaggt 9241 gccatctaag aagttcaagg ttctcggaaa caccgatagg cactctatca agaaaaacct 9301 tatcggtgct ctcctcttcg attctggtga aactgctgag gctaccagac tcaagagaac 9361 cgctagaaga aggtacacca gaagaaagaa caggatctgc tacctccaag agattttctc 9421 taacgagatg gctaaagtgg atgattcatt cttccacagg ctcgaagagt cattcctcgt 9481 ggaagaagat aagaagcacg agaggcaccc tatcttcgga aacatcgttg atgaggtggc 9541 ataccacgag aagtacccta ctatctacca cctcagaaag aagctcgttg attctactga 9601 taaggctgat ctcaggctca tctacctcgc tctcgctcac atgatcaagt tcagaggaca 9661 cttcctcatc gagggtgatc tcaaccctga taactctgat gtggataagt tgttcatcca 9721 gctcgtgcag acctacaacc agcttttcga agagaaccct atcaacgctt caggtgtgga 9781 tgctaaggct atcctctctg ctaggctctc taagtcaaga aggcttgaga acctcattgc 9841 tcagctccct ggtgagaaga agaacggact tttcggaaac ttgatcgctc tctctctcgg 9901 actcacccct aacttcaagt ctaacttcga tctcgctgag gatgcaaagc tccagctctc 9961 aaaggatacc tacgatgatg atctcgataa cctcctcgct cagatcggag atcagtacgc 10021 tgatttgttc ctcgctgcta agaacctctc tgatgctatc ctcctcagtg atatcctcag 10081 ggtgaacacc gagatcacca aggctccact ttctgcttct atgatcaaga gatacgatga 10141 gcaccaccag gatctcacac ttctcaaggc tcttgttaga cagcagctcc cagagaagta 10201 caaagaaatc ttcttcgatc agtctaagaa cggatacgct ggttacatcg atggtggtgc 10261 atctcaagaa gagttctaca agttcatcaa gccaatcttg gagaagatgg atggaaccga 10321 ggaactcctc gtgaagctca atagagagga tctccttagg aagcagagga ccttcgataa 10381 cggatctatc cctcatcaga tccacctcgg agagttgcac gctatcctta gaaggcaaga 10441 ggatttctac ccattcctca aggataacag agagaagatt gagaagatcc tcaccttcag 10501 aatcccttac tacgtgggac ctctcgctag aggaaactca agattcgctt ggatgaccag 10561 aaagtctgag gaaaccatca ccccttggaa cttcgaagag gtggtggata agggtgctag 10621 tgctcagtct ttcatcgaga ggatgaccaa cttcgataag aaccttccta acgagaaggt 10681 gctccctaag cactctttgc tctacgagta cttcaccgtg tacaacgagt tgaccaaggt 10741 taagtacgtg accgagggaa tgaggaagcc tgcttttttg tcaggtgagc aaaagaaggc 10801 tatcgttgat ctcttgttca agaccaacag aaaggtgacc gtgaagcagc tcaaagagga 10861 ttacttcaag aaaatcgagt gcttcgattc agtggaaatc tctggtgttg aggataggtt 10921 caacgcatct ctcggaacct accacgatct cctcaagatc attaaggata aggatttctt 10981 ggataacgag gaaaacgagg atatcttgga ggatatcgtt cttaccctca ccctcttcga 11041 ggatagagag atgatagaag aaaggctcaa gacctacgct catctcttcg atgataaggt 11101 gatgaagcag ttgaagagaa gaagatacac tggttgggga aggctctcaa gaaagctcat 11161 taacggaatc agggataagc agtctggaaa gacaatcctt gatttcctca agtctgatgg 11221 attcgctaac agaaacttca tgcagctcat ccacgatgat tctctcacct ttaaagagga 11281 tatccagaag gctcaggttt caggacaggg tgatagtctc catgagcata tcgctaacct 11341 cgctggatcc cctgcaatca agaagggaat cctccagact gtgaagattg tggatgagtt 11401 ggtgaaggtg atgggacaca agcctgagaa catcgtgatc gaaatggcta gagagaacca 11461 gaccactcag aagggacaga agaactctag ggaaaggatg aagaggatcg aggaaggtat 11521 caaagagctt ggatctcaga tcctcaaaga gcaccctgtt gagaacactc agctccagaa 11581 cgagaagctc tacctctact acttgcagaa cggaagggat atgtatgtgg atcaagagct 11641 tgatattaac aggctctctg attacgatgt tgatcatatc gtgccacagt cttttatcaa 11701 agatgattct atcgataaca aggtgctcac taggtctgat aagaacaggg gtaagagtga 11761 taacgtgcca agtgaagagg ttgtgaagaa aatgaagaac tattggaggc agctcctcaa 11821 cgctaagctc atcactcaga gaaagttcga taacttgacc aaggctgaga ggggaggact 11881 ctctgaattg gataaggcag gattcatcaa gagacagctc gtggaaacca ggcagatcac 11941 caaacatgtg gcacagatcc tcgattctag gatgaacacc aagtacgatg agaacgataa 12001 gttgatcagg gaagtgaagg ttatcaccct caagtcaaag ctcgtgtctg atttcagaaa 12061 ggatttccaa ttctacaagg tgagggaaat caacaactac caccacgctc acgatgctta 12121 ccttaacgct gttgttggaa ccgctctcat caagaagtat ccaaagttgg agtctgagtt 12181 cgtgtacggt gattataagg tgtacgatgt gaggaagatg atcgctaagt ctgagcaaga 12241 gatcggaaag gctaccgcta agtatttctt ctactctaac atcatgaatt tcttcaagac 12301 cgagatcact ctcgctaacg gtgagatcag aaagaggcca ctcatcgaga caaacggtga 12361 aacaggtgag atcgtgtggg ataagggaag ggatttcgct accgttagaa aggtgctctc 12421 tatgcctcag gtgaacatcg ttaagaaaac cgaggtgcag accggtggat tctctaaaga 12481 gtctatcctc cctaagagga actctgataa gctcattgct aggaagaagg attgggaccc 12541 taagaaatac ggtggtttcg attctcctac cgtggcttac tctgttctcg ttgtggctaa 12601 ggttgagaag ggaaagagta agaagctcaa gtctgttaag gaacttctcg gaatcactat 12661 catggaaagg tcatctttcg agaagaaccc aatcgatttc cttgaggcta agggatacaa 12721 agaggttaag aaggatctca tcatcaagct cccaaagtac tcacttttcg agttggagaa 12781 cggtagaaag aggatgctcg cttctgctgg tgagcttcaa aagggaaacg agcttgctct 12841 cccatctaag tacgttaact ttctttacct cgcttctcac tacgagaagt tgaagggatc 12901 tccagaagat aacgagcaga agcaactttt cgttgagcag cacaagcact acttggatga 12961 gatcatcgag cagatcagtg agttctctaa aagggtgatc ctcgctgatg caaacctcga 13021 taaggtgttg tctgcttaca acaagcacag agataagcct atcagggaac aggcagagaa 13081 catcatccat ctcttcaccc ttaccaacct cggtgctcct gctgctttca agtacttcga 13141 tacaaccatc gataggaaga gatacacctc taccaaagaa gtgctcgatg ctaccctcat 13201 ccatcagtct atcactggac tctacgagac taggatcgat ctctcacagc ttggaggtga 13261 tcctaagaag aaaagaaagg ttagatcttg atgacccggg tctccataat aatgtgtgag 13321 tagttcccag ataagggaat tagggttcct atagggtttc gctcatgtgt tgagcatata 13381 agaaaccctt agtatgtatt tgtatttgta aaatacttct atcaataaaa tttctaattc 13441 ctaaaaccaa aatccagtac taaaatccag atcccccgaa ttaaggcctt gacaggatat 13501 attggcgggt aaacctaaga gaaaagagcg tttattagaa taacggatat ttaaaactcg 13561 ag //