LOCUS LV01 12627 bp DNA circular 16-AUG-2017 DEFINITION modified cellbio126 from Gene Oracle. ACCESSION urn.local...1p-7x36jc9 KEYWORDS . SOURCE ORGANISM . FEATURES Location/Qualifiers misc_feature 6..223 /label="INT trunc" misc_feature 224..229 /Recognition_pattern="G^AATTC" /label="EcoRI" /note="Geneious type: restriction site" misc_feature 244..513 /label="U6" misc_feature 534..615 /label="gRNA scaffold" misc_feature 650..655 /Recognition_pattern="G^TCGAC" /label="SalI" /note="Geneious type: restriction site" misc_feature 668..675 /Recognition_pattern="TTAAT^TAA" /label="PacI" /note="Geneious type: restriction site" misc_feature 676..926 /label="trEF1a" misc_feature 927..933 /Recognition_pattern="A^CCWGGT" /label="SexAI" /note="Geneious type: restriction site" misc_feature 1009..1014 /Recognition_pattern="G^CTAGC" /label="NheI" /note="Geneious type: restriction site" misc_feature 1031..1624 /label="Puro" misc_feature 1625..1630 /Recognition_pattern="GGTAC^C" /label="KpnI" /note="Geneious type: restriction site" misc_feature 1631..1696 /label="2A" misc_feature 1712..5833 /label="Cas9" misc_feature 5834..5839 /Recognition_pattern="GTT^AAC" /label="HpaI" /note="Geneious type: restriction site" misc_feature 5840..5908 /label="2A" misc_feature 5909..5914 /Recognition_pattern="ATGCA^T" /label="NsiI" /note="Geneious type: restriction site" misc_feature 5915..6628 /label="GFP" misc_feature 6629..6634 /Recognition_pattern="GTT^AAC" /label="HpaI" /note="Geneious type: restriction site" misc_feature 6647..6652 /Recognition_pattern="T^CTAGA" /label="XbaI" /note="Geneious type: restriction site" polyA_signal 6668..7256 /label="WPRE" misc_feature 7271..7335 /label="Nef trunc" misc_feature 7319..7343 /label="PPT" misc_feature 7391..7486 /label="R(3')" misc_feature 7487..7569 /label="U5(3')" misc_feature 7956..7977 /label="T7 promoter" misc_feature 8144..8613 /label="f1 ori" misc_feature 8731..9591 /label="ampR" misc_feature 10772..10791 /label="T3 Promoter" /note="Geneious type: promoter prokaryotic" misc_structure 10816..11046 /label="RSV U3 frag" misc_structure 11047..11142 /label="R (5')" misc_structure 11143..11225 /label="U5 (5')" misc_signal 11288..11397 /label="Psi Element" misc_feature 11398..11719 /label="p17 trunc" misc_feature 11743..11830 /label="gp120 trunc" misc_binding 11831..12182 /label="RRE" misc_feature 12183..12597 /label="gp41 (trunc)" ORIGIN 1 tcgagcagtc gatcgagaca aatggcagta ttcatccaca attttaaaag aaaagggggg 61 attggggggt acagtgcagg ggaaagaata gtagacataa tagcaacaga catacaaact 121 aaagaattac aaaaacaaat tacaaaaatt caaaattttc gggtttatta cagggacagc 181 agagatccag tttggccgct ggatcgatgt acgggccaga tatgaattcg atatcgttta 241 aacacgcgta aggtcgggca ggaagagggc ctatttccca tgattccttc atatttgcat 301 atacgataca aggctgttag agagataatt agaattaatt tgactgtaaa cacaaagata 361 ttagtacaaa atacgtgacg tagaaagtaa taatttcttg ggtagtttgc agttttaaaa 421 ttatgtttta aaatggacta tcatatgctt accgtaactt gaaagtattt cgatttcttg 481 gctttatata tcttgtggaa aggacgaaac accgnnnnnn nnnnnnnnnn nnngttttag 541 agctagaaat agcaagttaa aataaggcta gtccgttatc aacttgaaaa agtggcaccg 601 agtcggtgct tttttctagg cccagctttc ttgtacaaag ttggcattag tcgacaagtg 661 agcaacctta attaatcgag tggctccggt gcccgtcagt gggcagagcg cacatcgccc 721 acagtccccg agaagttggg gggaggggtc ggcaattgaa ccggtgccta gagaaggtgg 781 cgcggggtaa actgggaaag tgatgtcgtg tactggctcc gcctttttcc cgagggtggg 841 ggagaaccgt atataagtgc agtagtcgcc gtgaacgttc tttttcgcaa cgggtttgcc 901 gccagaacac aggtgtcgtg acgcggacca ggttctggct aactagagaa cccactgctt 961 actggcttat cgaaattaat acgactcact atagggagac ccaagctggc tagccgccac 1021 catgaagctt accgagtaca agcccacggt gcgcctcgcc acccgcgacg acgtccccag 1081 ggccgtacgc accctcgccg ccgcgttcgc cgactacccc gccacgcgcc acaccgtcga 1141 tccggaccgc cacatcgagc gggtcaccga gctgcaagaa ctcttcctca cgcgcgtcgg 1201 gctcgacatc ggcaaggtgt gggtcgcgga cgacggcgcc gcggtggcgg tctggaccac 1261 gccggagagc gtcgaagcgg gggcggtgtt cgccgagatc ggcccgcgca tggccgagtt 1321 gagcggttcc cggctggccg cgcagcaaca gatggaaggc ctcctggcgc cgcaccggcc 1381 caaggagccc gcgtggttcc tggccaccgt cggcgtctcg cccgaccacc agggcaaggg 1441 tctgggcagc gccgtcgtgc tccccggagt ggaggcggcc gagcgcgccg gggtgcccgc 1501 cttcctggag acctccgcgc cccgcaacct ccccttctac gagcggctcg gcttcaccgt 1561 caccgccgac gtcgaggtgc ccgaaggacc gcgcacatgg tgcatgaccc gcaagcccgg 1621 tgccggtacc ggaagcggag ctactaactt cagcctgctg aagcaggctg gagacgtgga 1681 ggagaaccct ggacctaagc tttctcaatt gatggacaag aagtacagca tcggcctgga 1741 catcggcacc aactctgtgg gctgggccgt gatcaccgac gactacaagg tgcccagcaa 1801 gaaattcaag gtgctgggca acaccgaccg gcacagcatc aagaagaacc tgatcggcgc 1861 cctgctgttc ggctctggcg aaacagccga ggccacccgg ctgaagagaa ccgccagaag 1921 aagatacacc agacggaaga accggatctg ctatctgcaa gagatcttca gcaacgagat 1981 ggccaaggtg gacgacagct tcttccacag actggaagag tccttcctgg tggaagagga 2041 taagaagcac gagcggcacc ccatcttcgg caacatcgtg gacgaggtgg cctaccacga 2101 gaagtacccc accatctacc acctgagaaa gaagctggcc gacagcaccg acaaggccga 2161 cctgagactg atctacctgg ccctggccca catgatcaag ttccggggcc acttcctgat 2221 cgagggcgac ctgaaccccg acaacagcga cgtggacaag ctgttcatcc agctggtgca 2281 gatctacaat cagctgttcg aggaaaaccc catcaacgcc agcagagtgg acgccaaggc 2341 catcctgagc gccagactga gcaagagcag acggctggaa aatctgatcg cccagctgcc 2401 cggcgagaag cggaatggcc tgttcggcaa cctgattgcc ctgagcctgg gcctgacccc 2461 caacttcaag agcaacttcg acctggccga ggatgccaaa ctgcagctga gcaaggacac 2521 ctacgacgac gacctggaca acctgctggc ccagatcggc gaccagtacg ccgacctgtt 2581 tctggccgcc aagaacctgt ccgacgccat cctgctgagc gacatcctga gagtgaacag 2641 cgagatcacc aaggcccccc tgtccgcctc tatgatcaag agatacgacg agcaccacca 2701 ggacctgacc ctgctgaaag ctctcgtgcg gcagcagctg cctgagaagt acaaagagat 2761 tttcttcgac cagagcaaga acggctacgc cggctacatc gatggcggag ccagccagga 2821 agagttctac aagttcatca agcccatcct ggaaaagatg gacggcaccg aggaactgct 2881 cgtgaagctg aacagagagg acctgctgcg gaagcagcgg accttcgaca acggcagcat 2941 cccccaccag atccacctgg gagagctgca cgccattctg cggcggcagg aagattttta 3001 cccattcctg aaggacaacc gggaaaagat cgagaagatc ctgaccttca gaatccccta 3061 ctacgtgggc cctctggcca ggggaaacag cagattcgcc tggatgacca gaaagagcga 3121 ggaaaccatc accccctgga acttcgagga agtggtggac aagggcgcca gcgcccagag 3181 cttcatcgag cggatgacca acttcgataa gaacctgccc aacgagaagg tgctgcccaa 3241 gcacagcctg ctgtacgagt acttcaccgt gtacaacgag ctgaccaaag tgaaatacgt 3301 gaccgaggga atgcggaagc ccgcctttct gagcggcgag cagaaaaagg ccatcgtgga 3361 cctgctgttc aagaccaacc ggaaagtgac cgtgaagcag ctgaaagagg actacttcaa 3421 gaaaatcgag tgcttcgaca gcgtggaaat cagcggcgtg gaagatcggt tcaacgcctc 3481 cctgggcgcc tatcacgatc tgctgaaaat tatcaaggac aaggacttcc tggacaatga 3541 ggaaaacgag gacattctgg aagatatcgt gctgaccctg acactgtttg aggaccgggg 3601 catgatcgag gaacggctga aaacctatgc ccacctgttc gacgacaaag tgatgaagca 3661 gctgaagcgg cggagataca ccggctgggg caggctgagc cggaagctga tcaacggcat 3721 ccgggacaag cagtccggca agacaatcct ggatttcctg aagtccgacg gcttcgccaa 3781 cagaaacttc atgcagctga tccacgacga cagcctgacc tttaaagagg acatccagaa 3841 agcccaggtg tccggccagg gacactctct gcacgagcag atcgccaatc tggccggatc 3901 ccccgccatt aagaagggca tcctgcagac agtgaagatt gtggacgagc tcgtgaaagt 3961 gatgggccac aagcccgaga acatcgtgat cgaaatggcc agagagaacc agaccaccca 4021 gaagggacag aagaacagcc gcgagagaat gaagcggatc gaagagggca tcaaagagct 4081 gggcagccag atcctgaaag aacaccccgt ggaaaacacc cagctgcaga acgagaagct 4141 gtacctgtac tacctgcaga atgggcggga tatgtacgtg gaccaggaac tggacatcaa 4201 ccggctgtcc gactacgatg tggaccacat tgtgccccag tccttcatca aggacgactc 4261 catcgataac aaagtgctga ctcggagcga caagaaccgg ggcaagagcg acaacgtgcc 4321 ctccgaagag gtcgtgaaga agatgaagaa ctactggcgc cagctgctga atgccaagct 4381 gattacccag aggaagttcg acaatctgac caaggccgag agaggcggcc tgagcgaact 4441 ggataaggcc ggcttcatta agcggcagct ggtggaaacc cggcagatca caaagcacgt 4501 ggcacagatc ctggactccc ggatgaacac taagtacgac gagaacgaca aactgatccg 4561 ggaagtgaaa gtgatcaccc tgaagtccaa gctggtgtcc gacttcagaa aggatttcca 4621 gttttacaaa gtgcgcgaga tcaacaacta ccaccacgcc cacgacgcct acctgaacgc 4681 cgtcgtggga accgccctga tcaaaaagta ccctaagctg gaaagcgagt tcgtgtacgg 4741 cgattacaag gtgtacgacg tgcggaagat gatcgccaag agcgagcagg aaatcggcaa 4801 ggctaccgcc aagtacttct tctacagcaa catcatgaac tttttcaaga ccgagatcac 4861 actggccaac ggcgagatca gaaagcggcc tctgatcgag acaaacggcg aaaccgggga 4921 gatcgtgtgg gataagggcc gggattttgc cacagtgcgg aaagtgctgt ccatgcccca 4981 agtgaatatc gtgaaaaaga ccgaggtgca gaccggcggc ttcagcaaag agtctatcct 5041 gcccaagagg aactccgaca agctgatcgc cagaaagaag gattgggacc ctaagaagta 5101 cggcggcttt gacagcccca ccgtggccta ctctgtgctg gtggtggcca aagtggaaaa 5161 gggcaagtcc aagaaactga agagtgtgaa agagctgctg gggatcacca tcatggaaag 5221 aagcagcttc gagaagaatc ccatcgactt tctggaagcc aagggctaca aagaagtgaa 5281 aaaggacctg atcatcaagc tgcctaagta ctccctgttc gagctggaaa acggccggaa 5341 gcggatgctg gcttctgccg gcgaactgca gaagggaaac gagctggccc tgccctccaa 5401 atatgtgaac ttcctgtacc tggccagcca ctatgagaag ctgaagggct cccccgagga 5461 taatgagcag aaacagctgt ttgtggaaca gcacaagcac tacctggacg agatcatcga 5521 gcagattagc gagttctcca agcgcgtgat cctggccgat gccaacctgg acaaggtgct 5581 gagcgcctac aacaagcacc gggataagcc catcagagag caggccgaga atatcatcca 5641 cctgtttacc ctgaccaacc tgggagcccc tgccgccttc aagtactttg acaccaccat 5701 cgaccggaag aggtacacca gcaccaaaga ggtgctggac gccaccctga tccaccagag 5761 catcaccggc ctgtacgaga cacggatcga cctgtctcag ctgggaggcg accccaagaa 5821 aaagcgcaaa gtggttaacg gcagtggaga gggcagagga agtctgctaa catgcggtga 5881 cgtcgaggag aatcctggcc cacgatcgat gcatagcggg ggcgaggagc tgttcgccgg 5941 catcgtgccc gtgctgatcg agctggacgg cgacgtgcac ggccacaagt tcagcgtgcg 6001 cggcgagggc gagggcgacg ccgactacgg caagctggag atcaagttca tctgcaccac 6061 cggcaagctg cccgtgccct ggcccaccct ggtgaccacc ctctgctacg gcatccagtg 6121 cttcgcccgc taccccgagc acatgaagat gaacgacttc ttcaagagcg ccatgcccga 6181 gggctacatc caggagcgca ccatccagtt ccaggacgac ggcaagtaca agacccgcgg 6241 cgaggtgaag ttcgagggcg acaccctggt gaaccgcatc gagctgaagg gcaaggactt 6301 caaggaggac ggcaacatcc tgggccacaa gctggagtac agcttcaaca gccacaacgt 6361 gtacatccgc cccgacaagg ccaacaacgg cctggaggct aacttcaaga cccgccacaa 6421 catcgagggc ggcggcgtgc agctggccga ccactaccag accaacgtgc ccctgggcga 6481 cggccccgtg ctgatcccca tcaaccacta cctgagcact cagaccaaga tcagcaagga 6541 ccgcaacgag gcccgcgacc acatggtgct cctggagtcc ttcagcgcct gctgccacac 6601 ccacggcatg gacgagctgt acagggctgt taactgatga ctcgagtcta gaacgcgtta 6661 agtgcacaat caacctctgg attacaaaat ttgtgaaaga ttgactggta ttcttaacta 6721 tgttgctcct tttacgctat gtggatacgc tgctttaatg cctttgtatc atgctattgc 6781 ttcccgtatg gctttcattt tctcctcctt gtataaatcc tggttgctgt ctctttatga 6841 ggagttgtgg cccgttgtca ggcaacgtgg cgtggtgtgc actgtgtttg ctgacgcaac 6901 ccccactggt tggggcattg ccaccacctg tcagctcctt tccgggactt tcgctttccc 6961 cctccctatt gccacggcgg aactcatcgc cgcctgcctt gcccgctgct ggacaggggc 7021 tcggctgttg ggcactgaca attccgtggt gttgtcgggg aaatcatcgt cctttccttg 7081 gctgctcgcc tgtgttgcca cctggattct gcgcgggacg tccttctgct acgtcccttc 7141 ggccctcaat ccagcggacc ttccttcccc gggcctgctg ccggctctgc ggcctcttcc 7201 gcgtcttcgc cttcgccctc agacgagtcg gatctccctt tgggccgcct ccccgcccgc 7261 ggaacgtacc tttaagacca atgacttaca aggcagctgt agatcttagc cactttttaa 7321 aagaaaaggg gggactggaa gggctaattc actcccaacg aagacaagat ctgctttttg 7381 cttgtactgg gtctctctgg ttagaccaga tctgagcctg ggagctctct ggctaactag 7441 ggaacccact gcttaagcct caataaagct tgccttgagt gcttcaagta gtgtgtgccc 7501 gtctgttgtg tgactctggt aactagagat ccctcagacc cttttagtca gtgtggaaaa 7561 tctctagcag tagtagttca tgtcatctta ttattcagta tttataactt gcaaagaaat 7621 gaatatcaga gagtgagagg aacttgttta ttgcagctta taatggttac aaataaagca 7681 atagcatcac aaatttcaca aataaagcat ttttttcact gcattctagt tgtggtttgt 7741 ccaaactcat caatgtatct tatcatgtct ggctctagct atcccgcccc taactccgcc 7801 catcccgccc ctaactccgc ccagttccgc ccattctccg ccccatggct gactaatttt 7861 ttttatttat gcagaggccg aggccgcctc ggcctctgag ctattccaga agtagtgagg 7921 aggctttttt ggaggcctag ggacgtaccc aattcgccct atagtgagtc gtattacgcg 7981 cgctcactgg ccgtcgtttt acaacgtcgt gactgggaaa accctggcgt tacccaactt 8041 aatcgccttg cagcacatcc ccctttcgcc agctggcgta atagcgaaga ggcccgcacc 8101 gatcgccctt cccaacagtt gcgcagcctg aatggcgaat gggacgcgcc ctgtagcggc 8161 gcattaagcg cggcgggtgt ggtggttacg cgcagcgtga ccgctacact tgccagcgcc 8221 ctagcgcccg ctcctttcgc tttcttccct tcctttctcg ccacgttcgc cggctttccc 8281 cgtcaagctc taaatcgggg gctcccttta gggttccgat ttagtgcttt acggcacctc 8341 gaccccaaaa aacttgatta gggtgatggt tcacgtagtg ggccatcgcc ctgatagacg 8401 gtttttcgcc ctttgacgtt ggagtccacg ttctttaata gtggactctt gttccaaact 8461 ggaacaacac tcaaccctat ctcggtctat tcttttgatt tataagggat tttgccgatt 8521 tcggcctatt ggttaaaaaa tgagctgatt taacaaaaat ttaacgcgaa ttttaacaaa 8581 atattaacgc ttacaattta ggtggcactt ttcggggaaa tgtgcgcgga acccctattt 8641 gtttattttt ctaaatacat tcaaatatgt atccgctcat gagacaataa ccctgataaa 8701 tgcttcaata atattgaaaa aggaagagta tgagtattca acatttccgt gtcgccctta 8761 ttcccttttt tgcggcattt tgccttcctg tttttgctca cccagaaacg ctggtgaaag 8821 taaaagatgc tgaagatcag ttgggtgcac gagtgggtta catcgaactg gatctcaaca 8881 gcggtaagat ccttgagagt tttcgccccg aagaacgttt tccaatgatg agcactttta 8941 aagttctgct atgtggcgcg gtattatccc gtattgacgc cgggcaagag caactcggtc 9001 gccgcataca ctattctcag aatgacttgg ttgagtactc accagtcaca gaaaagcatc 9061 ttacggatgg catgacagta agagaattat gcagtgctgc cataaccatg agtgataaca 9121 ctgcggccaa cttacttctg acaacgatcg gaggaccgaa ggagctaacc gcttttttgc 9181 acaacatggg ggatcatgta actcgccttg atcgttggga accggagctg aatgaagcca 9241 taccaaacga cgagcgtgac accacgatgc ctgtagcaat ggcaacaacg ttgcgcaaac 9301 tattaactgg cgaactactt actctagctt cccggcaaca attaatagac tggatggagg 9361 cggataaagt tgcaggacca cttctgcgct cggcccttcc ggctggctgg tttattgctg 9421 ataaatctgg agccggtgag cgtgggtctc gcggtatcat tgcagcactg gggccagatg 9481 gtaagccctc ccgtatcgta gttatctaca cgacggggag tcaggcaact atggatgaac 9541 gaaatagaca gatcgctgag ataggtgcct cactgattaa gcattggtaa ctgtcagacc 9601 aagtttactc atatatactt tagattgatt taaaacttca tttttaattt aaaaggatct 9661 aggtgaagat cctttttgat aatctcatga ccaaaatccc ttaacgtgag ttttcgttcc 9721 actgagcgtc agaccccgta gaaaagatca aaggatcttc ttgagatcct ttttttctgc 9781 gcgtaatctg ctgcttgcaa acaaaaaaac caccgctacc agcggtggtt tgtttgccgg 9841 atcaagagct accaactctt tttccgaagg taactggctt cagcagagcg cagataccaa 9901 atactgttct tctagtgtag ccgtagttag gccaccactt caagaactct gtagcaccgc 9961 ctacatacct cgctctgcta atcctgttac cagtggctgc tgccagtggc gataagtcgt 10021 gtcttaccgg gttggactca agacgatagt taccggataa ggcgcagcgg tcgggctgaa 10081 cggggggttc gtgcacacag cccagcttgg agcgaacgac ctacaccgaa ctgagatacc 10141 tacagcgtga gctatgagaa agcgccacgc ttcccgaagg gagaaaggcg gacaggtatc 10201 cggtaagcgg cagggtcgga acaggagagc gcacgaggga gcttccaggg ggaaacgcct 10261 ggtatcttta tagtcctgtc gggtttcgcc acctctgact tgagcgtcga tttttgtgat 10321 gctcgtcagg ggggcggagc ctatggaaaa acgccagcaa cgcggccttt ttacggttcc 10381 tggccttttg ctggcctttt gctcacatgt tctttcctgc gttatcccct gattctgtgg 10441 ataaccgtat taccgccttt gagtgagctg ataccgctcg ccgcagccga acgaccgagc 10501 gcagcgagtc agtgagcgag gaagcggaag agcgcccaat acgcaaaccg cctctccccg 10561 cgcgttggcc gattcattaa tgcagctggc acgacaggtt tcccgactgg aaagcgggca 10621 gtgagcgcaa cgcaattaat gtgagttagc tcactcatta ggcaccccag gctttacact 10681 ttatgcttcc ggctcgtatg ttgtgtggaa ttgtgagcgg ataacaattt cacacaggaa 10741 acagctatga ccatgattac gccaagcgcg caattaaccc tcactaaagg gaacaaaagc 10801 tggagctgca agcttaatgt agtcttatgc aatactcttg tagtcttgca acatggtaac 10861 gatgagttag caacatgcct tacaaggaga gaaaaagcac cgtgcatgcc gattggtgga 10921 agtaaggtgg tacgatcgtg ccttattagg aaggcaacag acgggtctga catggattgg 10981 acgaaccact gaattgccgc attgcagaga tattgtattt aagtgcctag ctcgatacat 11041 aaacgggtct ctctggttag accagatctg agcctgggag ctctctggct aactagggaa 11101 cccactgctt aagcctcaat aaagcttgcc ttgagtgctt caagtagtgt gtgcccgtct 11161 gttgtgtgac tctggtaact agagatccct cagacccttt tagtcagtgt ggaaaatctc 11221 tagcagtggc gcccgaacag ggacttgaaa gcgaaaggga aaccagagga gctctctcga 11281 cgcaggactc ggcttgctga agcgcgcacg gcaagaggcg aggggcggcg actggtgagt 11341 acgccaaaaa ttttgactag cggaggctag aaggagagag atgggtgcga gagcgtcagt 11401 attaagcggg ggagaattag atcgcgatgg gaaaaaattc ggttaaggcc agggggaaag 11461 aaaaaatata aattaaaaca tatagtatgg gcaagcaggg agctagaacg attcgcagtt 11521 aatcctggcc tgttagaaac atcagaaggc tgtagacaaa tactgggaca gctacaacca 11581 tcccttcaga caggatcaga agaacttaga tcattatata atacagtagc aaccctctat 11641 tgtgtgcatc aaaggataga gataaaagac accaaggaag ctttagacaa gatagaggaa 11701 gagcaaaaca aaagtaagac caccgcacag caagcggccg ctgatcttca gacctggagg 11761 aggagatatg agggacaatt ggagaagtga attatataaa tataaagtag taaaaattga 11821 accattagga gtagcaccca ccaaggcaaa gagaagagtg gtgcagagag aaaaaagagc 11881 agtgggaata ggagctttgt tccttgggtt cttgggagca gcaggaagca ctatgggcgc 11941 agcgtcaatg acgctgacgg tacaggccag acaattattg tctggtatag tgcagcagca 12001 gaacaatttg ctgagggcta ttgaggcgca acagcatctg ttgcaactca cagtctgggg 12061 catcaagcag ctccaggcaa gaatcctggc tgtggaaaga tacctaaagg atcaacagct 12121 cctggggatt tggggttgct ctggaaaact catttgcacc actgctgtgc cttggaatgc 12181 tagttggagt aataaatctc tggaacagat ttggaatcac acgacctgga tggagtggga 12241 cagagaaatt aacaattaca caagcttaat acactcctta attgaagaat cgcaaaacca 12301 gcaagaaaag aatgaacaag aattattgga attagataaa tgggcaagtt tgtggaattg 12361 gtttaacata acaaattggc tgtggtatat aaaattattc ataatgatag taggaggctt 12421 ggtaggttta agaatagttt ttgctgtact ttctatagtg aatagagtta ggcagggata 12481 ttcaccatta tcgtttcaga cccacctccc aaccccgagg ggacccgaca ggcccgaagg 12541 aatagaagaa gaaggtggag agagagacag agacagatcc attcgattag tgaacggatc 12601 tcgacggtat cgatcacgag actagcc //