• USA Home
  • Search Results

  • tromethamine121147786111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tromethamine

4 Products
Synonym:   Tris
Image of Tromethamine
CAS Number:
Empirical Formula (Hill Notation):
 C4H11NO3
Molecular Weight:
 121.14
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Agency/Method
  Suitability
  Product Line
  Biological Source
  Special Grade
  Physical Form
Show More