• USA Home
  • Search Results

  • trishydroxymethylaminomethane121147786111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tris(hydroxymethyl)aminomethane

5 Products
Synonym:   2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol
Image of Tris(hydroxymethyl)<WBR>aminomethane
CAS Number:
Linear Formula:
 NH2C(CH2OH)3
Molecular Weight:
 121.14
 Beilstein/REAXYS Number:
 741883
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Physical Form
  Restriction
  Special Grade
  pH Value
  Vapor pressure
  Density
  Bulk Density
  Solubility
  Assay
  Potency
Show More