• USA Home
  • Search Results

  • trisbasemolecularbiologygradecas77861calbiochem121147786111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tris Base, Molecular Biology Grade - CAS 77-86-1 - Calbiochem

1 Products
Synonym:   2-Amino-2-(hydroxymethyl)-1,3-propanediol, tris(Hydroxymethyl)aminomethane, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol
Image of Tris Base, Molecular Biology Grade - CAS 77-86-1 - Calbiochem
CAS Number:
Linear Formula:
 NH2C(CH2OH)3
Molecular Weight:
 121.14
 Beilstein/REAXYS Number:
 741883
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Absorption
Show More