• USA Home
  • Search Results

  • omnipurtriscas77861calbiochem121147786111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

OmniPur TRIS -CAS 77-86-1 - Calbiochem

1 Products
Synonym:   2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(Hydroxymethyl)aminomethane, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Trizma, Tris(hydroxymethyl)aminomethane, Trometamol
Image of OmniPur<TMSYMBOL></TMSYMBOL> TRIS -CAS 77-86-1 - Calbiochem
CAS Number:
Linear Formula:
 NH2C(CH2OH)3
Molecular Weight:
 121.14
 Beilstein/REAXYS Number:
 741883
 MDL number:
 NACRES
 NA.21
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
Show More