• USA Home
  • Search Results

  • mopso225266839977911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

MOPSO

2 Products
Synonym:   β-Hydroxy-4-morpholinepropanesulfonic acid, 3-Morpholino-2-hydroxypropanesulfonic acid
Image of MOPSO
CAS Number:
Empirical Formula (Hill Notation):
 C7H15NO5S
Molecular Weight:
 225.26
 Beilstein/REAXYS Number:
 1109697
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Solubility
  Product Line
  Assay
Show More