• USA Home
  • Search Results

  • kanamycinsolutionfromstreptomyceskanamyceticus123452538994011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Kanamycin solution from Streptomyces kanamyceticus

2 Products
Image of Kanamycin solution from <I>Streptomyces kanamyceticus</I>
CAS Number:
 MDL number:
 NACRES:
 NA.76
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Mode of Action
  Physical Form
  Sterilization
  Product Line
  Concentration
  Biological Source
  Application(s)
Show More