• USA Home
  • Search Results

  • greensepbasicsfc3micronhplccolumn1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

GreenSep Basic SFC 3 Micron HPLC Column

12 Products
 NACRES
 SB.52
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Composite Dimension
  Application(s)
  Pore size
  Particle Dimension
  Matrix active group
  Manufacturer designation
Show More

Product #
Description
ES5118 GreenSep Basic SFC 3 Micron HPLC Column, 3 μm particle size, L × I.D. 5 cm × 2.1 mm,
ES5119 GreenSep Basic SFC 3 Micron HPLC Column, 3 μm particle size, L × I.D. 5 cm × 3.0 mm,
ES5120 GreenSep Basic SFC 3 Micron HPLC Column, 3 μm particle size, L × I.D. 5 cm × 4.6 mm,
ES5121 GreenSep Basic SFC 3 Micron HPLC Column, 3 μm particle size, L × I.D. 10 cm × 2.1 mm,
ES5122 GreenSep Basic SFC 3 Micron HPLC Column, 3 μm particle size, L × I.D. 10 cm × 3.0 mm,
ES5123 GreenSep Basic SFC 3 Micron HPLC Column, 3 μm particle size, L × I.D. 10 cm × 4.6 mm,
ES5124 GreenSep Basic SFC 3 Micron HPLC Column, 3 μm particle size, L × I.D. 15 cm × 2.1 mm,
ES5125 GreenSep Basic SFC 3 Micron HPLC Column, 3 μm particle size, L × I.D. 15 cm × 3.0 mm,
ES5126 GreenSep Basic SFC 3 Micron HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm,
ES5127 GreenSep Basic SFC 3 Micron HPLC Column, 3 μm particle size, L × I.D. 25 cm × 2.1 mm,
ES5128 GreenSep Basic SFC 3 Micron HPLC Column, 3 μm particle size, L × I.D. 25 cm × 3.0 mm,
ES5129 GreenSep Basic SFC 3 Micron HPLC Column, 3 μm particle size, L × I.D. 25 cm × 4.6 mm,