• USA Home
  • ES5126 - GreenSep Basic SFC 3 Micron HPLC Column

EMAIL THIS PAGE TO A FRIEND
ES5126

GreenSep Basic SFC 3 Micron HPLC Column

3 μm particle size, L × I.D. 15 cm × 4.6 mm

  •  NACRES SB.52

Purchase

Properties

Related Categories Analytical/Chromatography, GreenSep Basic 3 Micron SFC Columns, GreenSep Basic SFC HPLC Columns, HPLC/UHPLC, SFC (Supercritical Fluid Chromatography) Columns More...
Quality Level   100
mfr. no.   ES Industries, 135191-GS-BC
application(s)   HPLC: suitable
L × I.D.   15 cm × 4.6 mm
matrix active group   amine phase
particle size   3 μm
pore size   120 Å pore size
separation technique   supercritical fluid (SFC)
storage temp.   room temp

Description

General description

GreenSep Basic exhibits a highly basic character and is ideally suited to high performance/high speed SFC preparative chromatography applications of chemicals containing amine groups. The chromatogram shown below contains chemicals that are functionalized with amine and acidic groups and is a prime example of the superior peak shape performance and separation capacity obtainable with the GreenSep Basic column. It also compares GreenSep Basic with GreenSep Ethyl Pyridine showing the superior reten- tion on GreenSep Basic when compared to GreenSep Ethyl Pyridine.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

GreenSep is a trademark of ES Industries

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?