• USA Home
  • Z566640 - BudgetSensors AFM probes
EMAIL THIS PAGE TO A FRIEND
Z566640
BudgetSensors AFM probes
silicon tip, aluminum reflex coating, mode, contact
Purchase
Properties
Related Categories AFM Probes, Labware, Materials Science Labware More...
material   aluminum reflex coating
  silicon tip
feature   mode contact
mfr. no.   Innovative Solutions, ContAl-10
Description
General description
Useful for surface analysis in materials science, chemistry, physics and biology, and semiconductor applications.
• Silicon and silicon nitride probes
• Coatings: magnetic, gold, aluminum reflex, conductive chromium/platinum, gold/chromium
• Modes: soft, tapping, force modulation, contact
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?