• USA Home
  • CMC0013 - BIOTINYLATER F′ Electrocompetent Cells

EMAIL THIS PAGE TO A FRIEND
CMC0013 Sigma-Aldrich

BIOTINYLATER F′ Electrocompetent Cells

for production of biotinylated proteins

Purchase

Properties

grade   for molecular biology
cell transformation   competent cell type: electrocompetent
transformation efficiency: >1 x 1010 cfu/μg
shipped in   dry ice
storage temp.   −70°C

Description

General description

The BIOTINYLATER F′ Electrocompetent Cells are based on the MC1061 strain, but have had the birA gene integrated into the genome.
The birA protein (biotin ligase), which is induced with the addition of arabinose, will facilitate the addition of a biotin molecule to proteins bearing the appropriate signal sequence.

Genotype

MC1061 [F´ pro A+B+ lacIqZΔM15::Tn10 (TetR)] araD139 Δ(ara-leu)7696 Δ(lac)l74 galU galK hsdR2(rK- mK+) mcrB1 rpsL (StrR) birA

Features and Benefits

• Efficient in vivo biotinylation of target protein.
• Sole source of highly efficient electrocompetent cells (1 × 1010 cfu/μg).
• BirA gene stably integrated into genome for in vivo biotinylation of tagged proteins.

Components

• BIOTINYLATER F′ electrocompetent cells
• pUC 19 transformation control DNA
• recovery medium for cloning


Safety & Documentation

Safety Information

RIDADR 
UN 3245 9

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Competent Cell Selection & E. coli Markers Guide

We offer a range of Escherichia coli bacterial cells made competent with the highest efficiencies by optimized procedure specific to each strain. Choose from 24 new competent cells for a wide variety...
Keywords: Bacterial conjugations, Cloning, Gene expression, Methylations, Recombination, Transcription, transformation

Molecular Biology Handbook

Molecular Biology is the science that aims to understand biological activity at the molecular level. These biological activities usually involve the plant or animal cell, and the nucleic acids and pr...
Keywords: Cell culture, Cell signaling, Cloning, Detection methods, Electrophoresis, Gel electrophoresis, Gene expression, Genetics, Molecular biology, Molecular biology techniques, Nucleic acid hybridization, Nucleic acid purification, Polymerase chain reaction - quantitative, Protein electrophoresis, Purification, Transfection, Western blot, transformation

Protocols

Selecting Correctly Expressing Recombinants

Blue / White colony screening is a strategy to quickly and easily distinguish between recombinant and non-recombinant colonies. It requires a special vector and a special strain of E. coli. It is par...
Keywords: AGE, Amplification, Antibiotics, Cell disruption, Centrifugation, Cloning, Degradations, Detergents, Diagnostic, Digestions, Electrophoresis, Gel electrophoresis, Molecular biology, Peptide synthesis, Phase transitions, Polymerase chain reaction, Precipitation, Purification, Sequencing, Transfection, transformation

Transforming E. coli with Engineered Plasmid

Inoue and colleagues developed this method in 1990. It works well for many strains commonly used in cloning. The original method calls for growing the overnight E. coli cultures at 18°C. We find that...
Keywords: Antibiotics, Centrifugation, Cloning, Peptide synthesis, Phase transitions, transformation

Related Products

related product

Product #

Description

Add to Cart

L3022 LB Broth (Lennox), Highly-referenced microbial growth powder medium, low salt, suitable for salt-sensitive E.coli culture.
L2897 LB Broth with agar (Lennox), Highly-referenced microbial growth powder medium with Agar, low salt, suitable for salt-sensitive E. coli culture.
L5542 LB Agar Plates, pre-poured LB agar plates for E. coli
CMR0002 Recovery Medium for Cloning, for use with competent cells

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?