• USA Home
  • CMC0002 - SIG10 F′ Chemically Competent Cells

EMAIL THIS PAGE TO A FRIEND
CMC0002 Sigma-Aldrich

SIG10 F′ Chemically Competent Cells

for protein expression and DNA plasmid production

Purchase

Properties

Related Categories Cloning and Expression, Molecular Biology, Transformation of Bacteria More...
grade   for molecular biology
form   buffered aqueous solution
cell transformation   competent cell type: chemically competent
transformation efficiency: ≥5 x 108 cfu/μg
shipped in   dry ice
storage temp.   −70°C

Description

General description

An excellent choice for producing single-stranded DNA for sequencing, mutagenesis, etc.

Genotype

[F´ pro A+B+ lacIqZΔM15::Tn10 (TetR)] /mcrA Δ(mrr-hsdRMS-mcrBC) endA1 recA1 Φ80dlacZ?M15 ΔlacX74 araD139 Δ(ara, leu)7697 galU galK rpsL nupGλ tonA

Features and Benefits

• share the most useful genetic elements of standard cloning strains like DH5α, DH10B JM109, TOP10, etc. and directly replace them in cloning protocols.
• provide the performance researchers need with ease of use.
• provide solutions for a wide range of applications at economical prices.
• Blue - white screening

Components

• SIG10 F′ chemically competent cells
• pUC 19 transformation control DNA
• recovery medium for cloning

Legal Information

DH10B is a trademark of Life Technologies

DH5α is a trademark of Invitrogen Corp.

Safety & Documentation

Safety Information

RIDADR 
UN 3245 9

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Competent Cell Selection & E. coli Markers Guide

We offer a range of Escherichia coli bacterial cells made competent with the highest efficiencies by optimized procedure specific to each strain. Choose from 24 new competent cells for a wide variety...
Keywords: Bacterial conjugations, Cloning, Gene expression, Methylations, Recombination, Transcription, transformation

Molecular Biology Handbook

Molecular Biology is the science that aims to understand biological activity at the molecular level. These biological activities usually involve the plant or animal cell, and the nucleic acids and pr...
Keywords: Cell culture, Cell signaling, Cloning, Detection methods, Electrophoresis, Gel electrophoresis, Gene expression, Genetics, Molecular biology, Molecular biology techniques, Nucleic acid hybridization, Nucleic acid purification, Polymerase chain reaction - quantitative, Protein electrophoresis, Purification, Transfection, Western blot, transformation

Protocols

SIG10 Chemically Competent Cells

From our library of Protocols, Sigma-Aldrich presents SIG10 Chemically Competent Cells
Keywords: Antibiotics, Cloning, Peptide synthesis, transformation

Selecting Correctly Expressing Recombinants

Blue / White colony screening is a strategy to quickly and easily distinguish between recombinant and non-recombinant colonies. It requires a special vector and a special strain of E. coli. It is par...
Keywords: AGE, Amplification, Antibiotics, Cell disruption, Centrifugation, Cloning, Degradations, Detergents, Diagnostic, Digestions, Electrophoresis, Gel electrophoresis, Molecular biology, Peptide synthesis, Phase transitions, Polymerase chain reaction, Precipitation, Purification, Sequencing, Transfection, transformation

Transforming E. coli with Engineered Plasmid

Inoue and colleagues developed this method in 1990. It works well for many strains commonly used in cloning. The original method calls for growing the overnight E. coli cultures at 18°C. We find that...
Keywords: Antibiotics, Centrifugation, Cloning, Peptide synthesis, Phase transitions, transformation

Related Products

related product

Product #

Description

Add to Cart

L3022 LB Broth (Lennox), Highly-referenced microbial growth powder medium, low salt, suitable for salt-sensitive E.coli culture.
L2897 LB Broth with agar (Lennox), Highly-referenced microbial growth powder medium with Agar, low salt, suitable for salt-sensitive E. coli culture.
L5542 LB Agar Plates, pre-poured LB agar plates for E. coli
CMC0001 SIG10 Chemically Competent Cells, for protein expression and DNA plasmid production
CMC0007 SIG10 5a Chemically Competent Cells, for DNA plasmid production
CMR0002 Recovery Medium for Cloning, for use with competent cells

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?