HPLC
DiscoveryBIO Wide Pore C18
Discovery BIO Wide Pore C18
HPLC Columns and Capillaries
Bonded Phase:   Covalently-bonded octadecylsilane (C18), endcapped
Silica:   Spherical, 3µm, 5µm and 10µm diameters, 300Å pore diameter,
    high purity (<10ppm metals)
Solutions for Protein and Peptide Separation Challenges
  • Proteomics and biotherapeutics: high resolution of synthetic peptides and peptide maps
  • LC/MS; no-bleed, capillary dimensions, and pH stability
  • Peptide purification: Scalable from analytical to preparative
  • Reproducible separations batch-to-batch and column-to-column
 Here's the Proof
MATCHED SELECTIVITY FROM ANALYTICAL TO PREP
Column: Discovery BIO Wide Pore C18, 15cm x 4.6mm, 3µm; 15cm x 4.6mm, 5µm; 15cm x 10mm, 10µm, Mobile Phase: (A) 80:20, (0.1% TFA in H2O) : (0.1%TFA in MeCN), (B) 66:34, (0.1% TFA in H2O) : (0.1%TFA in MeCN), Flow Rate: 6.02cm/min, Temp: 30°C, Detection: 215nm, Sample: Peptide Mix (Cat. No. P2693)
Column Parameters & Run Conditions:
Column CV(mL) Injection(µL) Flow(mL/min)
15cm x 4.6mm, 3µm 1.64 5.0 1.00
15cm x 4.6mm, 5µm 1.71 5.2 1.00
15cm x 10mm, 10µm 8.01 24.5 4.73
Gradient:      
CV %A %B  
0 100 0  
2 100 0  
9 0 100  
PEPTIDE MAP OF CARBOXYMETHYLATED APOHEMOGLOBIN TRYPTIC DIGEST
Column: Discovery BIO Wide Pore C18, 15cm x 4.6mm, 5µm; Mobile Phase: (A) 95:5, (0.1% TFA in H2O):(0.1% TFA in MeCN); (B) 50:50, (0.1% TFA in H2O):(0.1% TFA in MeCN); Flow Rate: 1mL/min; Temp: 30°C; Detection: 215nm; Injection: 50µL; Sample: caboxymethylated apohemoglobin tryptic digest; Gradient: 0-100% B in 65 min
STABILITY OF DISCOVERY BIO WIDE PORE C18 AT ALKALINE pH
Alkaline pH Stability
Retention times of the sample components remained constant after 29,000 bed volumes were passed through the column.
Column: Discovery BIO Wide Pore C18, 5cm x 4.6mm, 5µm; Mobile Phase: 25mM potassium phosphate, pH 8 : MeOH (95:5); Flow Rate: 2mL/min; Temp: 35°C; Detection: 254nm
g001520
UNDETECTABLE LC/MS BLEED
LC/MS Bleed Analysis on Discovery BIO Wide Pore C18
Column: Discovery BIO Wide Pore C18, 15cm x 4.6mm, 3µm; Mobile Phase: (A) 0.1% TFA in H2O, (B) 0.1% TFA in MeOH; Flow Rate: 1mL/min; Temp: 30°C; Gradient: 0-100% B in 15 min, hold for 5 min, then back to 0% B for 10 min
No TIC baseline rise relative to blank run (gradient run w/o column)
LOT TO LOT REPRODUCIBILITY OF DISCOVERY BIO WIDE PORE C18
Column: Discovery BIO Wide Pore C18, 15cm x 4.6mm, 5µm; Mobile Phase: 80% 10mM ammonium acetate, pH 6.8 : 20% MeOH; Flow Rate: 1mL/min; Temp: 35°C; Detection: 254nm; Injection: 5µL
  1. Uracil (8µg/mL)
  2. Procainamide (30µg/mL)
  3. Sorbic acid (4µg/mL)
  4. Pyridine (100µg/mL)
  5. Caffeine (40µg/mL)
  6. Phenol (240µg/mL)
 Ordering Information
3µm Discovery BIO Wide Pore C18 HPLC Columns
ID(mm) Length(cm) Cat. No.
0.5 5 65517-U
2.1 5 567200-U
2.1 10 567201-U
2.1 15 567202-U
4.6 10 567204-U
4.6 15 567205-U
5µm Discovery BIO Wide Pore C18 HPLC Columns
ID(mm) Length(cm) Cat. No.
2.1 25 568203-U
4.0 25 568213-U
4.6 15 568222-U
4.6 25 568223-U
10.0 25 568230-U
10µm Discovery BIO Wide Pore C18 HPLC Columns
ID(mm) Length(cm) Cat. No.
10.0 5 567207-U
10.0 15 567208-U
10.0 25 567209-U
21.2 15 567211-U
21.2 25 567212-U
2cm Supelguard Cartridges with Discovery BIO C18 Packings
Description 2.1mm ID 4.0mm ID
3µm Particle Size
Kittriangle 567271-U 567273-U
5µm Particle Size
Kittriangle 568271-U 568273-U
For 4.0mm ID or 4.6mm ID analytical columns
triangle Kit includes a holder and one replaceable cartridge.