TraceCERT® and Certipur® Productportfolio and Product Features
TraceCERT® and Certipur® certified reference materials (CRMs) are developed and produced in an accredited laboratory, fulfilling both ISO/IEC 17025 and ISO 17034 guidelines. Our certificate of analysis includes the certification report and is issued according to the ISO Guide 31 guidelines.
|
AAS single solutions |
ICP single solutions |
|
TraceCERT® |
Certipur® |
TraceCERT® |
Certipur® |
Certification |
ISO/IEC 17025 and ISO 17034 |
Metal concentrations (mg/L) |
1’000 |
10’000
1’000
10
1 |
10’000
1’000
10 |
Matrix |
aqueous + different acid concentrations |
Packaging |
HD-PE bottle |
HD-PE bottle sealed in an
aluminum foil bag |
HD-PE bottle |
Package size |
250ml1,2 |
100ml, 500ml |
100ml1, 3 |
Indicative values |
|
Up to 70 impurities are tested whose concentrations are stated in the certificate |
1Hg (TraceCERT®) bottled in 100ml borosilicate white glass bottle
2 The following AAS (TraceCERT®) standards come in a 100ml package size: Pd, Au, Pt, Sc, Tl
The following AAS (Certipur®) standards come only in a 100ml package size: Cs, In, Te, Zr
3All 1mg/L ICP solutions as well as some special mixtures come in FEP bottles
Multi-element standard solutions |
|
TraceCERT® |
Certipur® |
Certification |
ISO/IEC 17025 and ISO 17034 |
|
Matrix |
Aqueous + various acid concentrations |
Packaging |
HD-PE bottle sealed in an aluminum foil bag |
HD-PE bottle |
Package size |
100ml |
100ml, 500ml* |
*Some tuning and calibration solutions come in a 500ml package size.
back to top
AAS and ICP Single Element Standards
(TraceCERT®and Certipur®)
We offer a large range of single element certified reference standard solutions in various concentrations (1 mg/L, 10 mg/L, 1’000 mg/L, 10’000 mg/L) suitable for AAS, ICP-OES or ICP-MS application.
To guarantee top reliability of the values, all certified reference solutions for AAS or ICP are traceable to specified NIST or BAM SRMs. We also show the NIST-traceable density on the Certificate of Analysis for our TraceCERT® line, together with information about the certification process, the traceability measurements, the storing behavior, the uncertainty evaluation and for the ICP product range the measured trace impurities.
AAS Single Element Standards, 1’000 mg/L
Element
|
Matrix
TraceCERT® |
Cat No
TraceCERT® |
Pck
TraceCERT® |
Matrix
Certipur® |
Cat No
Certipur® |
Pck
Certipur® |
Aluminum |
HNO3 |
39435 |
250ml |
HNO3 |
119770 |
100ml, 500ml |
Antimony |
HNO3 + HF traces |
94117 |
250ml |
HNO3 |
170204 |
100ml, 500ml |
Arsenic |
HNO3 + NaOH |
39436 |
250ml |
HNO3 |
119773 |
100ml, 500ml |
Barium |
HNO3 |
90092 |
250ml |
HNO3 |
119774 |
100ml, 500ml |
Beryllium |
HNO3 |
41856 |
250ml |
HNO3 |
170207 |
100ml, 500ml |
Bismuth |
HNO3 |
76668 |
250ml |
HNO3 |
119804 |
100ml, 500ml |
Boron |
H2O |
40591 |
250ml |
H2O |
119500 |
100ml, 500ml |
Cadmium |
HNO3 |
51994 |
250ml |
HNO3 |
119777 |
100ml, 500ml |
Calcium |
HNO3 |
69349 |
250ml |
HNO3 |
119778 |
100ml, 500ml |
Cerium |
HNO3 |
53378 |
250ml |
|
|
|
Cesium |
HNO3 |
67717 |
250ml |
HNO3 |
170212 |
100ml |
Chromium |
HNO3 |
02733 |
250ml |
HNO3 |
119779 |
100ml, 500ml |
Cobalt |
HNO3 |
05202 |
250ml |
HNO3 |
119785 |
100ml, 500ml |
Copper |
HNO3 |
38996 |
250ml |
HNO3 |
119786 |
100ml, 500ml |
Gallium |
HNO3 |
52874 |
100ml |
|
|
|
Germanium |
HNO3 |
92685 |
250ml |
|
|
|
Gold |
HCl |
08269 |
100ml |
HCl |
170216 |
100ml, 500ml |
Hafnium |
HNO3 + HF |
55816 |
100ml |
|
|
|
Indium |
HNO3 |
42225 |
100ml |
HNO3 |
119504 |
100ml |
Iron |
HNO3 |
16596 |
250ml |
HNO3 |
119781 |
100ml, 500ml |
Lanthanium |
HNO3 |
43678 |
100ml |
|
|
|
Lead |
HNO3 |
16595 |
250ml |
HNO3 |
119776 |
100ml, 500ml |
Lithium |
HNO3 |
59916 |
250ml |
HNO3 |
170223 |
100ml, 500ml |
Magnesium |
HNO3 |
42992 |
250ml |
HNO3 |
119788 |
100ml, 500ml |
Manganese |
HNO3 |
77036 |
250ml |
HNO3 |
119789 |
100ml, 500ml |
Mercury |
HNO3 |
16482 |
100ml* |
HNO3 |
170226 |
100ml, 500ml |
Molybdenum |
HCl |
67210 |
250ml |
H2O |
170227 |
100ml, 500ml |
Nickel |
HNO3 |
42242 |
250ml |
HNO3 |
119792 |
100ml, 500ml |
Niobium |
HNO3 + HF traces |
42887 |
100ml |
|
|
|
Palladium |
HCl |
78437 |
100ml |
HNO3 |
114282 |
100ml, 500ml |
Phosphorus |
H2O |
51474 |
250ml |
|
|
|
Platinum |
HCl + HNO3 traces |
47037 |
100ml |
HCl |
170219 |
100ml, 500ml |
Potassium |
HNO3 |
96665 |
250ml |
HNO3 |
170230 |
100ml, 500ml |
Rhenium |
HNO3 |
83787 |
100ml |
|
|
|
Rhodium |
HCl |
11561 |
100ml |
|
|
|
Rubidium |
HNO3 |
55727 |
100ml |
|
|
|
Ruthenium** |
HCl |
84033 |
100ml |
|
|
|
Scandium |
HNO3 |
68418 |
100ml |
HNO3 |
119513 |
100ml, 500ml |
Selenium |
HNO3 |
89498 |
250ml |
HNO3 |
119796 |
100ml, 500ml |
Silicon |
NaOH |
16259 |
250ml |
NaOH |
170236 |
100ml, 500ml |
Silver |
HNO3 |
39361 |
250ml |
HNO3 |
119797 |
100ml, 500ml |
Sodium |
HNO3 |
05201 |
250ml |
HNO3 |
170238 |
100ml, 500ml |
Strontium |
HNO3 |
51287 |
250ml |
HNO3 |
119799 |
100ml, 500ml |
Sulfur |
H2O |
18020 |
250ml |
|
|
|
Tellurium |
HNO3; <0.1% HF |
92027 |
250ml |
HNO3 |
119514 |
100ml |
Thallium |
HNO3 |
75159 |
100ml |
HNO3 |
119801 |
100ml, 500ml |
Tin |
HCl |
74244 |
250ml |
HCl |
170242 |
100ml, 500ml |
Titanium |
HNO3 |
04689 |
100ml |
H2O |
170243 |
100ml, 500ml |
Tungsten |
HNO3; 2% HF |
53465 |
100ml |
H2O |
170244 |
100ml, 500ml |
Vanadium |
HNO3 |
02334 |
250ml |
HNO3 |
170245 |
100ml, 500ml |
Yttrium |
HNO3 |
40423 |
250ml |
HNO3 |
119809 |
100ml, 500ml |
Zinc |
HNO3 |
18827 |
250ml |
HNO3 |
119806 |
100ml, 500ml |
Zirconium |
HNO3; 0.2% HF |
73574 |
250ml |
HCl |
170234 |
100ml |
*in borosilicate glass bottle
** no primary reference material available
back to top
ICP Single Element Standards, 1mg/L
Element |
Matrix TraceCERT® |
Cat No TraceCERT® |
Pck TraceCERT® |
Aluminum |
HNO3 |
05674 |
100ml |
Antimony |
HNO3; <0.2% HF |
89519 |
100ml |
Arsenic |
HNO3 |
75016 |
100ml |
Barium |
HNO3 |
75017 |
100ml |
Boron |
H2O |
41511 |
100ml |
Cadmium |
HNO3 |
12313 |
100ml |
Calcium |
HNO3 |
41599 |
100ml |
Chromium (III) |
HNO3 |
72995 |
100ml |
Cobalt |
HNO3 |
41798 |
100ml |
Copper |
HNO3 |
41621 |
100ml |
Gold |
HCl + HNO3 traces |
67363 |
100ml |
Indium |
HNO3 |
56875 |
100ml |
Iron |
HNO3 |
68436 |
100ml |
Lead |
HNO3 |
75015 |
100ml |
Lithium |
HNO3 |
39185 |
100ml |
Magnesium |
HNO3 |
53906 |
100ml |
Manganese |
HNO3 |
42058 |
100ml |
Molybdenum |
H2O |
04488 |
100ml |
Nickel |
HNO3 |
72631 |
100ml |
Palladium |
HNO3 + HCl traces |
42396 |
100ml |
Phosphorus |
H2O |
93674 |
100ml |
Platinum |
HCl + HNO3 traces |
49763 |
100ml |
Potassium |
HNO3 |
30937 |
100ml |
Scandium |
HNO3 |
67464 |
100ml |
Selenium |
HNO3 |
56544 |
100ml |
Silver |
HNO3 |
69389 |
100ml |
Sodium |
HNO3 |
80831 |
100ml |
Strontium |
HNO3 |
96672 |
100ml |
Sulfur |
H2O |
68356 |
100ml |
Tin |
HNO3, <0.1% HF |
63564 |
100ml |
Titanium |
HNO3, <0.1% HF |
44266 |
100ml |
Vanadium |
HNO3 |
75593 |
100ml |
Yttrium |
HNO3 |
75592 |
100ml |
Zinc |
HNO3 |
75594 |
100ml |
back to top
ICP Single Element Standards, 10 mg/L
Element |
Matrix
TraceCERT® |
Cat No
TraceCERT® |
Pck
TraceCERT® |
Matrix
Certipur® |
Cat No
Certipur® |
Pck Certipur® |
Rhodium |
|
|
|
HNO3 |
108525 |
100ml |
Thorium |
|
|
|
HNO3 |
170391 |
100ml |
Uranium |
|
|
|
HNO3 |
170360 |
100ml |
Mercury |
HNO3 |
89459 |
100ml |
|
|
|
ICP Single Element Standards, 1’000 mg/L
** no primary reference standard available
ICP Single Element Standards, 10’000 mg/L
back to top
Titrisol® Concentrated Single-Element Standards for AAS
We offer an extensive range of Titrisol® concentrated single-element standards. Simply dilute the concentrate in order to produce the desired standard solution. Titrisol® concentrates are traceable to standard reference material from NIST and are provided in easy-to-handle plastic ampoules. Unopened Titrisol® ampoules have a minimum expiration time of 5 years. A comprehensive batch-specific Certificate of Analysis can be downloaded from our webpage, using product and lot number.
Dilution of Titrisol® concentrates
In preparing a dilution series, the concentrates should be diluted with the same concentration of acid as is in the standard itself. Titrisol® concentrates should be diluted to 1l with 0.1 mol/l HCl in the first step. Alternatively, 30ml of 30% HCl can be placed in a volumetric flask and made up to 1l with distilled water. Further dilutions should then be made with distilled water.
Minimum expiration time |
Unopened Titrisol® ampoules |
5 years |
Solution of 1’000 mg/l |
Recommended max. 12 months |
Solution of 10-100 mg/l |
Recommended 1 month |
Solution of 1-10 mg/l |
Recommended max 1 week |
Solution of <1 mg/l |
Recommended 1 day |
As exceptions, the following Titrisol® concentrates should be diluted as indicated: |
Gold |
Place 500ml 30% hydrochloric acid in a volumetric flask and make up to 1l with distilled water. |
Lead, Selenium, Silver |
Dilute to 1 l with 0.1 mol/L or place 5ml 65% nitric acid in a volumetric flask and make up to the mark. |
Molybdenum |
Place 10ml 25% ammonia solution in a volumetric flask and make up to 1 l with distilled water. |
Silicon |
Make up to the mark with distilled water |
Vanadium |
Place 5ml 95% sulfuric acid in a volumetric flask and make up to 1 l with distilled water. |
Product range
back to top
Multi-element Standards for ICP – TraceCERT® and Certipur® solutions
TraceCERT® multi-element solutions
Our TraceCERT® multielement solutions are certified reference materials (CRMs), produced and certified under
ISO/IEC 17025 and ISO 17034. All CRMs are traceable to primary material from a National Metrology Institute (NMI).
Periodic Table Mixes
With these three multielement standard solutions, all ICP measurable elements of the periodic table are covered (excluding mercury). All components are present at a concentration of 10 mg/L.
|
|
92091 |
41135 |
78334 |
|
|
Periodic table mix 1 for ICP |
Periodic table mix 2 for ICP |
Periodic table mix 3 for ICP |
|
|
In 10% HNO3 |
In 5% HCl + 1% HF |
In 5% HNO3 |
All concentrations are in mg/L |
Aluminum |
Al |
10 |
|
|
Antimony |
Sb |
|
10 |
|
Arsenic |
As |
10 |
|
|
Barium |
Ba |
10 |
|
|
Beryllium |
Be |
10 |
|
|
Bismuth |
Bi |
10 |
|
|
Boron |
B |
10 |
|
|
Cadmium |
Cd |
10 |
|
|
Calcium |
Ca |
10 |
|
|
Cesium |
Cs |
10 |
|
|
Chromium |
Cr |
10 |
|
|
Cobalt |
Co |
10 |
|
|
Copper |
Cu |
10 |
|
|
Gallium |
Ga |
10 |
|
|
Germanium |
Ge |
|
10 |
|
Gold |
Au |
|
10 |
|
Hafnium |
Hf |
|
10 |
|
Indium |
In |
10 |
|
|
Iridium |
Ir |
|
10 |
|
Iron |
Fe |
10 |
|
|
Lanthanum |
La |
|
|
10 |
Lead |
Pb |
10 |
|
|
Lithium |
Li |
10 |
|
|
Magnesium |
Mg |
10 |
|
|
Manganese |
Mn |
10 |
|
|
Mercury |
Hg |
|
|
|
Molybdenum |
Mo |
|
10 |
|
Nickel |
Ni |
10 |
|
|
Niobium |
Nb |
|
10 |
|
Palladium |
Pd |
|
10 |
|
Phosphorus |
P |
10 |
|
|
Platinum |
Pt |
|
10 |
|
Potassium |
K |
10 |
|
|
Rhenium |
Re |
|
10 |
|
Rhodium |
Rh |
|
10 |
|
Rubidium |
Rb |
10 |
|
|
Ruthenium |
Ru |
|
10 |
|
Scandium |
Sc |
|
|
10 |
Selenium |
Se |
10 |
|
|
Silicon |
Si |
10 |
|
|
Silver |
Ag |
10 |
|
|
Sodium |
Na |
10 |
|
|
Strontium |
Sr |
10 |
|
|
Sulfur |
S |
10 |
|
|
Tantalum |
Ta |
|
10 |
|
Tellurium |
Te |
10 |
|
|
Thallium |
Tl |
10 |
|
|
Tin |
Sn |
|
10 |
|
Titanium |
Ti |
|
10 |
|
Tungsten |
W |
|
10 |
|
Vanadium |
V |
10 |
|
|
Yttrium |
Y |
|
|
10 |
Zinc |
Zn |
10 |
|
|
Zirconium |
Zr |
|
10 |
|
Pck size |
|
100ml |
100ml |
100ml |
Chemical Group Mixes
Each of these eight multielement standard solutions contains a specific group of elements. Concentrations of the analytes are 100 mg/L except for the rare elements mix where the analytes have a concentration of 50 mg/L.
|
|
96441 |
67288 |
67349 |
55263 |
51509 |
04330 |
96442 |
93468 |
|
|
Alkali Metal Mix |
Alkaline Earth Metal Mix |
Rare Earth
Metal Mix |
Metalloid & Non-Metal Mix |
Post-transition Metal Mix |
Transition
Metal Mix 1 |
Transition
Metal Mix 2 |
Transition
Metal Mix 3 |
|
|
In 2% HNO3 |
In 2% HNO3 |
In 2% HNO3 |
In 10% HNO3 + <0.5% HF |
In 2% HNO3 |
In 2% HNO3 |
In 2% HNO3 + 1% HF |
In 5% HNO3 |
All concentrations are in mg/L |
Aluminum |
Al |
|
100 |
|
|
|
Antimony |
Sb |
|
|
|
100 |
|
Arsenic |
As |
|
|
|
100 |
|
Barium |
Ba |
|
100 |
|
Beryllium |
Be |
|
100 |
|
Bismuth |
Bi |
|
100 |
|
|
|
Boron |
B |
|
|
|
100 |
|
Cadmium |
Cd |
|
100 |
|
|
Calcium |
Ca |
|
100 |
|
Cerium |
Ce |
|
|
50 |
|
Cesium |
Cs |
100 |
|
Chromium |
Cr |
|
100 |
|
|
Cobalt |
Co |
|
100 |
|
|
Copper |
Cu |
|
100 |
|
|
Dysprosium |
Dy |
|
|
50 |
|
Erbium |
Er |
|
|
50 |
|
Europium |
Eu |
|
|
50 |
|
Gadolinium |
Gd |
|
|
50 |
|
Gallium |
Ga |
|
100 |
|
|
|
Germanium |
Ge |
|
|
|
100 |
|
Gold |
Au |
|
100 |
Hafnium |
Hf |
|
100 |
|
Holmium |
Ho |
|
|
50 |
|
Indium |
In |
|
100 |
|
|
|
Iridium |
Ir |
|
100 |
Iron |
Fe |
|
100 |
|
|
Lanthanum |
La |
|
|
50 |
|
Lead |
Pb |
|
100 |
|
|
|
Lithium |
Li |
100 |
|
Lutetium |
Lu |
|
|
50 |
|
Magnesium |
Mg |
|
100 |
|
Manganese |
Mn |
|
100 |
|
|
Mercury |
Hg |
|
Molybdenum |
Mo |
|
100 |
|
Neodymium |
Nd |
|
|
50 |
|
Nickel |
Ni |
|
100 |
|
|
Niobium |
Nb |
|
100 |
|
Osmium |
Os |
|
100 |
Palladium |
Pd |
|
100 |
Phosphorus |
P |
|
|
|
100 |
|
Platinum |
Pt |
|
100 |
Potassium |
K |
100 |
|
Praseodymium |
Pr |
|
|
50 |
|
Rhenium |
Re |
|
100 |
|
Rhodium |
Rh |
|
100 |
Rubidium |
Rb |
100 |
|
Ruthenium |
Ru |
|
100 |
Samarium |
Sm |
|
|
50 |
|
Scandium |
Sc |
|
|
50 |
|
Selenium |
Se |
|
|
|
100 |
|
Silicon |
Si |
|
|
|
100 |
|
Silver |
Ag |
|
100 |
|
|
Sodium |
Na |
100 |
|
Strontium |
Sr |
|
100 |
|
Sulfur |
S |
|
|
|
100 |
|
Tantalum |
Ta |
|
100 |
|
Tellurium |
Te |
|
|
|
100 |
|
Terbium |
Tb |
|
|
50 |
|
Thallium |
Tl |
|
100 |
|
|
|
Thulium |
Tm |
|
|
50 |
|
Tin |
Sn |
|
|
|
100 |
|
Titanium |
Ti |
|
100 |
|
Tungsten |
W |
|
100 |
|
Vanadium |
V |
|
100 |
|
|
Ytterbium |
Yb |
|
|
50 |
|
Yttrium |
Y |
|
|
50 |
|
Zinc |
Zn |
|
100 |
|
|
Zirconium |
Zr |
|
100 |
|
Pck size |
|
100ml |
100ml |
100ml |
100ml |
100ml |
100ml |
100ml |
100ml |
Other TraceCERT® multi-element solutions
|
90243 |
49596 |
51844 |
54704 |
43843 |
91529 |
Multi-element standard solutions |
1 for ICP |
3 for ICP |
4 for ICP |
5 for ICP |
6 for ICP |
Metal mix I for ICP |
All concentrations are in mg/L |
|
In 10% HNO3 |
In 5% HNO3 |
In 10% HNO3 |
In 10% HNO3 |
In 5% HNO3; <0.1% HF |
In 4.8% HNO3 + 0.5% HF |
Aluminum |
Al |
50 |
|
40 |
10 |
100 |
100 |
Antimony |
Sb |
|
100 |
|
Arsenic |
As |
|
|
40 |
|
|
|
Barium |
Ba |
10 |
|
40 |
10 |
100 |
|
Beryllium |
Be |
|
|
10 |
10 |
|
|
Bismuth |
Bi |
100 |
|
|
10 |
|
|
Boron |
B |
50 |
|
100 |
|
100 |
100 |
Cadmium |
Cd |
10 |
|
10 |
10 |
100 |
|
Calcium |
Ca |
|
2000 |
|
10 |
100 |
100 |
Cesium |
Cs |
|
|
|
10 |
|
|
Chromium |
Cr |
50 |
|
20 |
10 |
100 |
|
Cobalt |
Co |
10 |
|
10 |
10 |
100 |
|
Copper |
Cu |
10 |
|
20 |
10 |
100 |
|
Gallium |
Ga |
|
|
|
10 |
|
|
Indium |
In |
|
|
|
10 |
|
|
Iron |
Fe |
10 |
|
100 |
100 |
100 |
100 |
Lead |
Pb |
100 |
|
40 |
10 |
100 |
|
Lithium |
Li |
50 |
|
|
10 |
100 |
|
Magnesium |
Mg |
10 |
400 |
|
10 |
100 |
100 |
Manganese |
Mn |
10 |
|
10 |
10 |
100 |
100 |
Mercury |
Hg |
|
100 |
|
Molybdenum |
Mo |
50 |
|
|
10 |
|
|
Nickel |
Ni |
50 |
|
20 |
10 |
100 |
|
Phosphorus |
P |
|
100 |
|
Potassium |
K |
100 |
200 |
|
100 |
100 |
100 |
Rubidium |
Rb |
|
|
|
10 |
|
|
Selenium |
Se |
|
|
100 |
|
|
|
Silicon |
Si |
|
100 |
|
Silver |
Ag |
10 |
|
|
10 |
|
|
Sodium |
Na |
50 |
1000 |
|
100 |
100 |
100 |
Strontium |
Sr |
10 |
|
|
10 |
|
|
Thallium |
Tl |
50 |
|
100 |
10 |
|
|
Titanium |
Ti |
|
100 |
|
Tungsten |
W |
|
100 |
Vanadium |
V |
|
|
40 |
10 |
100 |
|
Zinc |
Zn |
10 |
|
100 |
10 |
100 |
100 |
|
Pck size |
|
100ml |
100ml |
100ml |
100ml |
100ml |
100ml |
back to top
Certipur® portfolio
Our Certipur® multi-element calibration solutions are traceable to standard reference material from NIST. A certificate of analysis is enclosed in each package. It includes data on content, composition, traceability, date of release and expiry date.
|
1.15474 |
1.11355 |
1.09492 |
1.09494 |
1.09493 |
1.09491 |
1.09480 |
1.09487 |
1.09495 |
Multi-element standard solutions |
I |
IV |
VIII |
IX |
X |
XI |
XIII |
XVI |
VII |
application |
|
|
|
toxic elements, Cr (VI) |
For surface water |
For sewage sludge |
|
1 mol/L HNO3 |
1 mol/L HNO3 |
1 mol/L HNO3 |
1 mol/L HNO3 |
1 mol/L HNO3 |
1 mol/L HNO3 |
5% HNO3 |
5% HNO3 |
15% HCl |
All concentrations are in mg/L |
Aluminum |
Al |
100 |
1000 |
100 |
|
|
|
500 |
|
|
Antimony |
Sb |
|
100 |
100 |
Arsenic |
As |
|
|
|
100 |
50 µg/L
|
|
100 |
100 |
|
Barium |
Ba |
5 |
1000 |
100 |
|
50 µg/L
|
Beryllium |
Be |
1 |
|
100 |
100 |
20 µg/L
|
|
100 |
100 |
|
Bismuth |
Bi |
200 |
1000 |
100 |
|
10 µg/L
|
Boron |
B |
15 |
1000 |
100 |
|
100 µg/L
|
Cadmium |
Cd |
20 |
1000 |
100 |
100 |
20 µg/L
|
10 |
25 |
100 |
|
Calcium |
Ca |
|
1000 |
100 |
|
35000 µg/L
|
|
|
100 |
|
Chromium |
Cr |
25 |
1000 |
100 |
100 |
20
µg/L |
900 |
100 |
100 |
|
Cobalt |
Co |
20 |
1000 |
100 |
|
25 µg/L
|
|
100 |
100 |
|
Copper |
Cu |
20 |
1000 |
100 |
|
20 µg/L
|
800 |
100 |
100 |
|
Gallium |
Ga |
150 |
1000 |
100 |
|
Hafnium |
Hf |
|
100 |
Indium |
In |
200 |
1000 |
|
Iridium |
Ir |
|
100 |
Iron |
Fe |
15 |
1000 |
100 |
|
100 µg/L
|
|
100 |
100 |
|
Lead |
Pb |
200 |
1000 |
100 |
100 |
25 µg/L
|
900 |
100 |
100 |
|
Lithium |
Li |
|
1000 |
100 |
|
100 |
|
Magnesium |
Mg |
|
1000 |
100 |
|
15000 µg/L
|
|
|
100 |
|
Manganese |
Mn |
5 |
1000 |
100 |
|
30 µg/L
|
8 |
100 |
100 |
|
Mercury |
Hg |
|
|
|
100 |
|
|
5 |
|
|
Molybdenum |
Mo |
|
100 µg/L
|
Nickel |
Ni |
50 |
1000 |
100 |
100 |
50 µg/L
|
200 |
100 |
100 |
|
Phosphorus |
P |
|
100 |
|
Potassium |
K |
|
1000 |
100 |
|
3000 µg/L
|
Selenium |
Se |
|
|
100 |
100 |
10 µg/L
|
|
25 |
100 |
|
Silver |
Ag |
50 |
1000 |
|
Sodium |
Na |
|
1000 |
100 |
|
8000 µg/L
|
Strontium |
Sr |
1 |
1000 |
100 |
|
100 µg/L
|
|
|
100 |
|
Tantalum |
Ta |
|
100 |
Tellurium |
Te |
|
|
100 |
|
Thallium |
Tl |
400 |
1000 |
100 |
100 |
10 µg/L
|
|
|
100 |
|
Tin |
Sn |
|
100 |
Titanium |
Ti |
|
100 |
100 |
Vanadium |
V |
|
50 µg/L
|
|
250 |
100 |
|
Zinc |
Zn |
20 |
1000 |
100 |
|
50 µg/L
|
2500 |
100 |
100 |
|
Zirconium |
Zr |
|
100 |
|
Pck size |
|
100ml |
100ml |
100ml |
100ml |
100ml |
100ml |
100ml |
100ml |
100ml |
|
1.10580 |
1.09411 |
1.09410 |
1.09500 |
1.10714 |
1.09481 |
Multi-element standard solutions |
VI |
XXIV |
XXIII |
XVIII |
V |
XIV |
application |
calibration in ICP-MS |
Tuning solution 700 ES |
For mass calibration |
For calibration |
HCl soluble elements |
HCl soluble elements |
|
1 mol/L HNO3 |
1% HNO3 |
5% HNO3 |
5% HNO3 |
5% HCl |
2% HCl |
Pck size |
100ml |
500ml |
500ml |
100ml |
500ml |
500ml |
All concentrations are in mg/L |
Aluminum |
Al |
10 |
50 |
|
100 |
20 |
|
Antimony |
Sb |
|
|
|
100 |
|
|
Arsenic |
As |
100 |
50 |
|
100 |
20 |
20 |
Barium |
Ba |
10 |
50 |
1 |
50 |
2 |
|
Beryllium |
Be |
100 |
|
|
5 |
1 |
|
Boron |
B |
100 |
|
1 |
50 |
2 |
|
Cadmium |
Cd |
10 |
50 |
|
|
2 |
|
Calcium |
Ca |
1000 |
|
|
|
10 |
|
Chromium |
Cr |
10 |
50 |
|
20 |
2 |
|
Cobalt |
Co |
10 |
50 |
1 |
50 |
2 |
|
Copper |
Cu |
10 |
50 |
|
50 |
|
|
Gallium |
Ga |
10 |
|
1 |
|
|
|
Indium |
In |
|
|
1 |
|
|
|
Iron |
Fe |
100 |
|
1 |
20 |
2 |
20 |
Lead |
Pb |
10 |
50 |
|
100 |
20 |
20 |
Lithium |
Li |
10 |
|
1 |
|
2 |
|
Lutetium |
Lu |
|
|
1 |
|
|
|
Magnesium |
Mg |
10 |
50 |
|
20 |
1 |
|
Manganese |
Mn |
10 |
|
|
|
1 |
20 |
Mercury |
Hg |
|
5 |
|
Molybdenum |
Mo |
10 |
50 |
|
|
|
20 |
Nickel |
Ni |
10 |
50 |
|
50 |
5 |
20 |
Phosphorus |
P |
|
10 |
100 |
Potassium |
K |
10 |
500 |
1 |
|
100 |
100 |
Rhodium |
Rh |
|
|
1 |
|
|
|
Rubidium |
Rb |
10 |
|
Scandium |
Sc |
|
|
1 |
|
1 |
20 |
Selenium |
Se |
100 |
50 |
|
100 |
20 |
|
Silver |
Ag |
10 |
|
|
10 |
|
|
Sodium |
Na |
10 |
|
1 |
|
20 |
20 |
Strontium |
Sr |
10 |
50 |
|
|
1 |
|
Sulfur |
S |
|
100 |
Tellurium |
Te |
10 |
|
|
|
20 |
|
Thallium |
Tl |
10 |
|
1 |
100 |
2 |
|
Uranium |
U |
10 |
|
1 |
|
|
|
Vanadium |
V |
10 |
|
Yttrium |
Y |
|
|
1 |
|
1 |
|
Zinc |
Zn |
100 |
50 |
|
|
2 |
|
|
Pck size |
|
100ml |
100ml |
100ml |
100ml |
100ml |
100ml |
back to top
TraceCERT® mixes for special applications
Mixes acc. to ICHQ3D guidelines (pharma testing)
We offer different Inorganic elemental impurity mixes as certified reference material standard mixes with element ratios corresponding to the oral, parenteral and inhalation elemental concentrations limits of the ICHQ3D guideline.
Mixes for cannabis testing
It is critical to accurately monitor levels of elemental impurities in cannabis products, both to ensure regulatory compliance – and more importantly – consumer and patient safety. Trace levels of heavy metals such as arsenic, cadmium, mercury and lead may accumulate in plant material through uptake from soil and, more commonly, may be introduced through the use of certain fertilizers.
Our heavy metal mixes are each tailored to specific US state specific regulations and offer concentrations that match the specific regulatory bodies‘ action and reporting limits.
We also offer mixes with flat concentrations.
All TraceCERT® mixes are certified reference materials, certified according to ISO/IEC 17025 and ISO 17034.
You can find more information about this specific product group on our Cannabis Testing webpage